Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02103737_u1
          See other LOC689766 GE Assays ›
          Gene Symbol:
          LOC689766
          Gene Name:
          hypothetical protein LOC689766
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 95885184 - 95908393 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02103737_u1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC689766
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109549.1 NP_001103019.1 4 465 159
          XM_006250332.2 XP_006250394.1 4 500 159
          XM_008769801.1 XP_008768023.1 2 591 159

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000864.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Amplicon spans exons and probe does not span exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003491 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003489 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68559 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072455 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003491 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003491 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072455 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003491 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072455 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003489 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766040 - -
          Affymetrix Rat Gene 1.0 ST Array 10766043 - -
          Affymetrix Rat Gene 1.0 ST Array 10766041 - -
          Affymetrix Rat Gene 1.0 ST Array 10766042 - -
          Affymetrix Rat Gene 1.1 ST Array 10766039 - -
          Affymetrix Rat Clariom S Assay PSR1300006087.rn.1 TC1300000864.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006085.rn.1 TC1300000864.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006088.rn.1 TC1300000864.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006086.rn.1 TC1300000864.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006089.rn.1 TC1300000864.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006090.rn.1 TC1300000864.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300006084.rn.1 TC1300000864.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10766039 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20744938 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0