Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02104329_s1
          See other TSHZ1 GE Assays ›
          Gene Symbol:
          Tshz1
          Gene Name:
          teashirt zinc finger homeobox 1
          Gene Aliases:
          Sdccag33
          Chromosome Location:
          Chr.18: 80860995 - 80907744 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02104329_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tshz1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008772238.1 XP_008770460.1 3 1431 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          anterior/posterior pattern specification
          regulation of gene expression
          middle ear morphogenesis
          soft palate development
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P514987 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P514987 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P514987 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P514987 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042975 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P514987 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P526795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805513 - -
          Rat Genome 230 2.0 Array 1392611_at - -
          Rat Genome 230 2.0 Array 1380283_at - -
          Rat Genome 230 2.0 Array 1395149_at - -
          Rat Genome 230 2.0 Array 1383573_at - -
          Rat Genome 230 2.0 Array 1383573_PM_at - -
          Rat Genome 230 2.0 Array 1392611_PM_at - -
          Rat Genome 230 2.0 Array 1395149_PM_at - -
          Rat Genome 230 2.0 Array 1380283_PM_at - -
          Affymetrix HT Rat Focus Array 1380283_PM_at - -
          Affymetrix HT Rat Focus Array 1383573_PM_at - -
          Affymetrix HT Rat Focus Array 1383573_at - -
          Affymetrix HT Rat Focus Array 1380283_at - -
          Rat Expression Set 230 1395149_at - -
          Rat Expression Set 230 1380283_at - -
          Rat Expression Set 230 1380283_PM_at - -
          Rat Expression Set 230 1383573_PM_at - -
          Rat Expression Set 230 1392611_PM_at - -
          Rat Expression Set 230 1392611_at - -
          Rat Expression Set 230 1383573_at - -
          Rat Expression Set 230 1395149_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22394087 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368226 - -
          RatRef-12 Expression BeadChip ILMN_58517 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0