Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02114356_s1
          See other SLC19A3 GE Assays ›
          Gene Symbol:
          Slc19a3
          Gene Name:
          solute carrier family 19 (thiamine transporter), member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 88764073 - 88828412 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02114356_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc19a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108228.1 NP_001101698.1 8 1609 167

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Transporter 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          drug transmembrane transport
          thiamine transmembrane transport
          thiamine uptake transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P008166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170677 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119857 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008166 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008166 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P008166 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121983 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94802 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929455 - -
          Affymetrix Rat Gene 1.0 ST Array 10929452 - -
          Affymetrix Rat Gene 1.0 ST Array 10929454 - -
          Affymetrix Rat Gene 1.0 ST Array 10929453 - -
          Affymetrix Rat Gene 1.0 ST Array 10929451 - -
          Affymetrix Rat Gene 1.1 ST Array 10929450 - -
          Affymetrix Rat Clariom S Assay PSR0900019278.rn.1 TC0900002347.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019266.rn.1 TC0900002347.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019268.rn.1 TC0900002347.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019281.rn.1 TC0900002347.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019282.rn.1 TC0900002347.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019277.rn.1 TC0900002347.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019267.rn.1 TC0900002347.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900019279.rn.1 TC0900002347.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10929450 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21684773 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352427 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline