Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02345875_m1
          See other TMEM206 GE Assays ›
          Gene Symbol:
          Tmem206
          Gene Name:
          transmembrane protein 206
          Gene Aliases:
          RGD1359339
          Chromosome Location:
          Chr.13: 110006975 - 110032790 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02345875_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem206
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007679.1 NP_001007680.1 7 - 8 960 141
          XM_006250481.2 XP_006250543.1 6 - 7 784 141
          XM_008769855.1 XP_008768077.1 7 - 8 992 141
          GenBank mRNA BC082061.1 - 7 - 8 960 7132483 141
          FQ225543.1 - 7 - 8 969 141
          FQ228821.1 - 8 - 9 1002 141
          FQ233431.1 - 7 - 8 965 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001048.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem206

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P682216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7771 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15127 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101535 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P682216 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P682216 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P682216 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P682216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99081 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41357 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766732 - -
          Affymetrix Rat Gene 1.0 ST Array 10766731 - -
          Affymetrix Rat Gene 1.0 ST Array 10766729 - -
          Affymetrix Rat Gene 1.0 ST Array 10766728 - -
          Affymetrix Rat Gene 1.0 ST Array 10766733 - -
          Affymetrix Rat Gene 1.0 ST Array 10766727 - -
          Affymetrix Rat Gene 1.0 ST Array 10766730 - -
          Affymetrix Rat Gene 1.0 ST Array 10766734 - -
          Affymetrix Rat Gene 1.1 ST Array 10766726 - -
          Rat Genome 230 2.0 Array 1393165_at - -
          Rat Genome 230 2.0 Array 1393165_PM_at - -
          Affymetrix HT Rat Focus Array 1393165_at - -
          Affymetrix HT Rat Focus Array 1393165_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300007488.rn.1 TC1300001048.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300007489.rn.1 TC1300001048.rn.1 RTA TC
          Rat Expression Set 230 1393165_PM_at - -
          Rat Expression Set 230 1393165_at - -
          Affymetrix Rat Gene 1.0 ST Array 10766726 - -
          Rat Genome U34 Set rc_AI029600_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350631 - -
          RatRef-12 Expression BeadChip ILMN_51236 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0