Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02346972_m1
          See other ZFP868 GE Assays ›
          Gene Symbol:
          Zfp868
          Gene Name:
          zinc finger protein 868
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 21510794 - 21525290 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02346972_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp868
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009538.1 NP_001009538.1 4 - 5 585 110
          XM_006252915.2 XP_006252977.1 5 - 6 742 110
          XM_006252916.2 XP_006252978.1 5 - 6 953 110
          XM_006252917.2 XP_006252979.1 4 - 5 878 110
          GenBank mRNA BC062078.1 - 4 - 5 585 5599629 110
          BC089094.1 - 4 - 5 608 7125789 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000327.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp868

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P20885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55680 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20885 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20885 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P20885 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20885 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1054708 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56531 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10787716 - -
          Affymetrix Rat Gene 1.0 ST Array 10787712 - -
          Affymetrix Rat Gene 1.0 ST Array 10787715 - -
          Affymetrix Rat Gene 1.0 ST Array 10787713 - -
          Affymetrix Rat Gene 1.0 ST Array 10787711 - -
          Affymetrix Rat Gene 1.0 ST Array 10787717 - -
          Affymetrix Rat Gene 1.1 ST Array 10787710 - -
          Rat Genome 230 2.0 Array 1382113_at - -
          Rat Genome 230 2.0 Array 1382113_PM_at - -
          Affymetrix HT Rat Focus Array 1382113_at - -
          Affymetrix HT Rat Focus Array 1382113_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600003454.rn.1 TC1600000327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600003455.rn.1 TC1600000327.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600003456.rn.1 TC1600000327.rn.1 RTA TC
          Rat Expression Set 230 1382113_PM_at - -
          Rat Expression Set 230 1382113_at - -
          Affymetrix Rat Gene 1.0 ST Array 10787710 - -
          Rat Genome U34 Set rc_AI178530_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21432208 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373226 - -
          RatRef-12 Expression BeadChip ILMN_67653 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline