Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02531655_s1
          See other OLR1493 GE Assays ›
          Gene Symbol:
          Olr1493
          Gene Name:
          olfactory receptor gene Olr1493
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 60455040 - 60455966 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02531655_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olr1493
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006609.1 NP_001006610.1 889 84
          GenBank mRNA M64387.1 - 889 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olr1493

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P154162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83956 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152364 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21083 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117126 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31919 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99238 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154162 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154162 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P154162 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410516 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55859 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81354 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97133 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47967 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99555 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52393 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54292 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89660 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87434 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34462 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11553 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80531 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90309 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735741 - -
          Affymetrix Rat Gene 1.0 ST Array 10735740 - -
          Affymetrix Rat Gene 1.1 ST Array 10735737 - -
          Rat Genome 230 2.0 Array 1388092_at - -
          Rat Genome 230 2.0 Array 1388092_PM_at - -
          Affymetrix HT Rat Focus Array 1388092_at - -
          Affymetrix HT Rat Focus Array 1388092_PM_at - -
          Rat Expression Set 230 1388092_at - -
          Rat Expression Set 230 1388092_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735737 - -
          Rat Genome U34 Set AF091572_s_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline