Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02532420_g1
          See other ATP5G3 GE Assays ›
          Gene Symbol:
          Atp5g3
          Gene Name:
          ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C3 (subunit 9)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 60811218 - 60813899 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02532420_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp5g3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053756.1 NP_446208.1 4 661 76
          XM_006234393.2 XP_006234455.1 5 690 76
          XM_006234394.2 XP_006234456.1 5 670 76
          GenBank mRNA AF315374.1 - 4 661 76
          FQ217141.1 - 5 628 76
          FQ224352.1 - 4 596 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp5g3

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          aging
          ATP synthesis coupled proton transport
          ATP hydrolysis coupled proton transport
          response to ethanol
          molecular_function
          lipid binding
          hydrogen ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15344 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P484950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422780 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15344 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P484950 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15344 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P484950 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15344 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P484950 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15344 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P484950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24736 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846250 - -
          Affymetrix Rat Gene 1.0 ST Array 10846251 - -
          Affymetrix Rat Gene 1.0 ST Array 10846249 - -
          Affymetrix Rat Gene 1.0 ST Array 10940472 - -
          Affymetrix Rat Gene 1.1 ST Array 10940472 - -
          Affymetrix Rat Gene 1.1 ST Array 10846248 - -
          Rat Genome 230 2.0 Array 1367620_at - -
          Rat Genome 230 2.0 Array 1367620_PM_at - -
          Affymetrix HT Rat Focus Array 1367620_at - -
          Affymetrix HT Rat Focus Array 1367620_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300025901.rn.1 TC0300003334.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025896.rn.1 TC0300003334.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025904.rn.1 TC0300003334.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025902.rn.1 TC0300003334.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300025899.rn.1 TC0300003334.rn.1 RTA TC
          Rat Expression Set 230 1367620_at - -
          Rat Expression Set 230 1367620_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846248 - -
          Affymetrix Rat Gene 1.0 ST Array 10940472 - -
          Rat Genome U34 Set rc_AI180425_at - -
          Rat Genome U34 Set rc_AI102979_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22155345 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354415 - -
          RatRef-12 Expression BeadChip ILMN_52419 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline