Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02586190_s1
          See other TRIL GE Assays ›
          Gene Symbol:
          Tril
          Gene Name:
          TLR4 interactor with leucine-rich repeats
          Gene Aliases:
          RGD1310827
          Chromosome Location:
          Chr.4: 83967695 - 83972540 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02586190_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tril
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034010.1 NP_001029182.1 1 4739 80
          GenBank mRNA BC100659.1 - 1 4739 7935751 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tril

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine production involved in immune response
          inflammatory response
          toll-like receptor 4 signaling pathway
          innate immune response
          lipopolysaccharide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P423566 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859349 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18141 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P423566 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158564 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P423566 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158564 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P423566 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158564 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P286236 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P423574 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P423566 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102056 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862608 - -
          Affymetrix Rat Gene 1.0 ST Array 10862606 - -
          Affymetrix Rat Gene 1.0 ST Array 10862607 - -
          Affymetrix Rat Gene 1.1 ST Array 10862605 - -
          Rat Genome 230 2.0 Array 1374496_at - -
          Rat Genome 230 2.0 Array 1374496_PM_at - -
          Affymetrix HT Rat Focus Array 1374496_at - -
          Affymetrix HT Rat Focus Array 1374496_PM_at - -
          Rat Expression Set 230 1374496_PM_at - -
          Rat Expression Set 230 1374496_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862605 - -
          Rat Genome U34 Set rc_AI228167_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21021436 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369348 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline