Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03416394_m1
          See other GIMD1 GE Assays ›
          Gene Symbol:
          Gimd1
          Gene Name:
          GIMAP family P-loop NTPase domain containing 1
          Gene Aliases:
          RGD1563706
          Chromosome Location:
          Chr.2: 237679681 - 237714434 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03416394_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gimd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113782.1 NP_001107254.1 2 - 3 682 71
          XM_006233317.2 XP_006233379.1 2 - 3 690 71
          XM_008761516.1 XP_008759738.1 2 - 3 482 71
          GenBank mRNA BC158622.1 - 2 - 3 682 7454123 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002231.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Process(es)

          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P21956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175054 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84346 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P21956 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P21956 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36106 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46346 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92703 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0200015344.rn.1 TC0200002231.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015351.rn.1 TC0200002231.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015352.rn.1 TC0200002231.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015350.rn.1 TC0200002231.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015353.rn.1 TC0200002231.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20717489 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372584 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline