Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03810003_s1
          See other PNMA3 GE Assays ›
          Gene Symbol:
          Pnma3
          Gene Name:
          paraneoplastic Ma antigen 3
          Gene Aliases:
          RGD1563752
          Chromosome Location:
          Chr.X: 153063307 - 153067500 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03810003_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pnma3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106342.2 NP_001099812.1 2 1352 67
          XM_006229513.2 XP_006229575.1 2 1800 67
          XM_006229514.2 XP_006229576.1 3 1800 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001258.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Amplicon spans exons and probe does not span exons

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P403302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256166 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P403302 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P403302 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P403302 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P403302 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935871 - -
          Affymetrix Rat Gene 1.0 ST Array 10935870 - -
          Affymetrix Rat Gene 1.0 ST Array 10935872 - -
          Affymetrix Rat Gene 1.1 ST Array 10935867 - -
          Rat Genome 230 2.0 Array 1381360_PM_at - -
          Rat Genome 230 2.0 Array 1381360_at - -
          Affymetrix Rat Clariom S Assay PSR0X00007524.rn.1 TC0X00001258.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007526.rn.1 TC0X00001258.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007525.rn.1 TC0X00001258.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007518.rn.1 TC0X00001258.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007523.rn.1 TC0X00001258.rn.1 RTA TC
          Rat Expression Set 230 1381360_at - -
          Rat Expression Set 230 1381360_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10935867 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359267 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline