Search
Search
| Gene Symbol: Mir93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031853.1 | 1 | 48 | 61 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-25 | Mir25 | rno-mir-25 | -120.0 | Sense | Chr. 12 - 19307340 - 19307423 on NCBI Build Rnor_6.0 |
| Downstream | microRNA mir-106b | Mir106b | rno-mir-106b | 123.0 | Sense | Chr. 12 - 19307752 - 19307833 on NCBI Build Rnor_6.0 |
Set Membership: |
|