Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GAGGCAGCCAGGTCTGGGAACTTCA[C/T]CCCCAACTGCGGTCCTGGCCAGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO9 - anoctamin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012302.2 | Intron | NP_001012302.2 | ||||
XM_011520048.1 | Intron | XP_011518350.1 | ||||
XM_011520051.2 | Intron | XP_011518353.1 | ||||
XM_011520052.2 | Intron | XP_011518354.1 | ||||
XM_011520053.2 | Intron | XP_011518355.1 | ||||
XM_017017644.1 | Intron | XP_016873133.1 | ||||
XM_017017645.1 | Intron | XP_016873134.1 | ||||
XM_017017646.1 | Intron | XP_016873135.1 | ||||
XM_017017647.1 | Intron | XP_016873136.1 |
SIGIRR - single Ig and TIR domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |