Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AAATTGCTCCTTTGATTAGGAGATG[A/G]TTGCTACAGTTTGCAACGGAATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TLR5 - toll like receptor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003268.5 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | NP_003259.2 | |
XM_005273241.4 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | XP_005273298.2 | |
XM_005273242.4 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | XP_005273299.2 | |
XM_005273243.4 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | XP_005273300.2 | |
XM_006711504.3 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | XP_006711567.1 | |
XM_006711505.3 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | XP_006711568.1 | |
XM_006711506.3 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | XP_006711569.1 | |
XM_011509937.2 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | XP_011508239.1 | |
XM_017002208.1 | 3060 | Missense Mutation | ACC,ATC | T,I 856 | XP_016857697.1 |