Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TTGATTGGCTTGTCAATGAGGATTG[A/G]CTTTGCCAATTATGTTGTGTAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIG1 - androgen induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286587.1 | Intron | NP_001273516.1 | ||||
NM_001286588.1 | Intron | NP_001273517.1 | ||||
NM_001286589.1 | Intron | NP_001273518.1 | ||||
NM_016108.3 | Intron | NP_057192.2 | ||||
XM_011535881.2 | Intron | XP_011534183.1 | ||||
XM_011535882.2 | Intron | XP_011534184.1 | ||||
XM_011535883.2 | Intron | XP_011534185.1 | ||||
XM_011535884.2 | Intron | XP_011534186.1 | ||||
XM_011535885.2 | Intron | XP_011534187.1 | ||||
XM_017010927.1 | Intron | XP_016866416.1 | ||||
XM_017010928.1 | Intron | XP_016866417.1 |