Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
ATCTCCGGAGCGAGGAACAGAAGAA[G/T]AAGAACCGGAAGAAACTGTCCGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610323 | ||||||||||||||||||||
Literature Links: |
MTDH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTDH - metadherin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178812.3 | 883 | Missense Mutation | AAG,AAT | K,N 116 | NP_848927.2 | |
XM_005251099.3 | 883 | Missense Mutation | AAG,AAT | K,N 116 | XP_005251156.1 | |
XM_011517367.2 | 883 | Missense Mutation | AAG,AAT | K,N 116 | XP_011515669.1 | |
XM_011517368.2 | 883 | Missense Mutation | AAG,AAT | K,N 116 | XP_011515670.1 | |
XM_011517369.2 | 883 | Missense Mutation | AAG,AAT | K,N 116 | XP_011515671.1 | |
XM_011517370.2 | 883 | Missense Mutation | AAG,AAT | K,N 116 | XP_011515672.1 | |
XM_017013966.1 | 883 | Missense Mutation | AAG,AAT | K,N 116 | XP_016869455.1 | |
XM_017013967.1 | 883 | Missense Mutation | AAG,AAT | K,N 116 | XP_016869456.1 | |
XM_017013968.1 | 883 | Missense Mutation | AAG,AAT | K,N 116 | XP_016869457.1 |