Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TCATCATTTTCTTCCTCCTCCTCTA[C/T]CTGGTCTGGCAAGACCTCTTTCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300964 | ||||||||||||||||||||
Literature Links: |
LAS1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAS1L - LAS1 like, ribosome biogenesis factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170649.1 | 1760 | Missense Mutation | ATA,GTA | I,V 562 | NP_001164120.1 | |
NM_001170650.1 | 1760 | Missense Mutation | ATA,GTA | I,V 520 | NP_001164121.1 | |
NM_031206.4 | 1760 | Missense Mutation | ATA,GTA | I,V 579 | NP_112483.1 | |
XM_005262301.1 | 1760 | Missense Mutation | ATA,GTA | I,V 579 | XP_005262358.1 | |
XM_005262304.1 | 1760 | Intron | XP_005262361.1 | |||
XM_005262305.3 | 1760 | Intron | XP_005262362.1 | |||
XM_005262306.3 | 1760 | Intron | XP_005262363.1 | |||
XM_011531045.1 | 1760 | Missense Mutation | ATA,GTA | I,V 537 | XP_011529347.1 | |
XM_011531046.1 | 1760 | Intron | XP_011529348.1 | |||
XM_017029877.1 | 1760 | Missense Mutation | ATA,GTA | I,V 562 | XP_016885366.1 | |
XM_017029878.1 | 1760 | Intron | XP_016885367.1 | |||
XM_017029879.1 | 1760 | Missense Mutation | ATA,GTA | I,V 277 | XP_016885368.1 | |
XM_017029880.1 | 1760 | Missense Mutation | ATA,GTA | I,V 260 | XP_016885369.1 |