Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_362932780_10
          See other HPDL GT Assays ›
          SNP ID:
          rs759295912
          Gene
          HPDL MUTYH TOE1
          Gene Name
          4-hydroxyphenylpyruvate dioxygenase like
          mutY DNA glycosylase
          target of EGR1, member 1 (nuclear)
          Set Membership:
          -
          Chromosome Location:
          Chr.1: 45332464 - 45332464 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          CGGACACGGCACAGCACCCGTGCTA[C/T]GTTGCCATCCACCACACCGGTTGCC

          Assay ID C_362932780_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          2 submissions

          Phenotype:

          MIM: 604933 MIM: 613931

          Literature Links:

          HPDL PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          HPDL - 4-hydroxyphenylpyruvate dioxygenase like
          There are no transcripts associated with this gene.
          MUTYH - mutY DNA glycosylase
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001048171.1 683 Missense Mutation ATA,GTA I,V 225 NP_001041636.1
          NM_001048172.1 683 Missense Mutation ATA,GTA I,V 212 NP_001041637.1
          NM_001048173.1 683 Missense Mutation ATA,GTA I,V 211 NP_001041638.1
          NM_001048174.1 683 Missense Mutation ATA,GTA I,V 211 NP_001041639.1
          NM_001128425.1 683 Missense Mutation ATA,GTA I,V 239 NP_001121897.1
          NM_001293190.1 683 Missense Mutation ATA,GTA I,V 226 NP_001280119.1
          NM_001293191.1 683 Missense Mutation ATA,GTA I,V 222 NP_001280120.1
          NM_001293192.1 683 Missense Mutation ATA,GTA I,V 119 NP_001280121.1
          NM_001293195.1 683 Missense Mutation ATA,GTA I,V 211 NP_001280124.1
          NM_001293196.1 683 Missense Mutation ATA,GTA I,V 119 NP_001280125.1
          NM_012222.2 683 Missense Mutation ATA,GTA I,V 236 NP_036354.1
          XM_011541497.2 683 Missense Mutation ATA,GTA I,V 231 XP_011539799.1
          XM_011541498.1 683 Missense Mutation ATA,GTA I,V 225 XP_011539800.1
          XM_011541499.1 683 Missense Mutation ATA,GTA I,V 225 XP_011539801.1
          XM_011541500.2 683 Missense Mutation ATA,GTA I,V 225 XP_011539802.1
          XM_011541501.2 683 Missense Mutation ATA,GTA I,V 225 XP_011539803.1
          XM_011541502.2 683 Missense Mutation ATA,GTA I,V 225 XP_011539804.1
          XM_011541503.2 683 Missense Mutation ATA,GTA I,V 225 XP_011539805.1
          XM_011541504.2 683 Missense Mutation ATA,GTA I,V 222 XP_011539806.1
          XM_011541505.2 683 Missense Mutation ATA,GTA I,V 85 XP_011539807.1
          XM_011541506.1 683 Missense Mutation ATA,GTA I,V 85 XP_011539808.1
          XM_011541507.2 683 Missense Mutation ATA,GTA I,V 73 XP_011539809.2
          XM_017001331.1 683 Missense Mutation ATA,GTA I,V 225 XP_016856820.1
          XM_017001332.1 683 Missense Mutation ATA,GTA I,V 225 XP_016856821.1
          XM_017001333.1 683 Missense Mutation ATA,GTA I,V 225 XP_016856822.1
          XM_017001334.1 683 Missense Mutation ATA,GTA I,V 212 XP_016856823.1
          XM_017001335.1 683 Missense Mutation ATA,GTA I,V 119 XP_016856824.1
          XM_017001336.1 683 Missense Mutation ATA,GTA I,V 96 XP_016856825.1
          XM_017001337.1 683 Missense Mutation ATA,GTA I,V 96 XP_016856826.1
          XM_017001338.1 683 Missense Mutation ATA,GTA I,V 96 XP_016856827.1
          XM_017001339.1 683 Missense Mutation ATA,GTA I,V 96 XP_016856828.1
          TOE1 - target of EGR1, member 1 (nuclear)
          There are no transcripts associated with this gene.

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA glycosylase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          mismatch repair
          response to oxidative stress
          depurination
          DNA binding
          protein binding
          DNA N-glycosylase activity
          MutLalpha complex binding
          MutLbeta complex binding
          MutSalpha complex binding
          MutSbeta complex binding
          metal ion binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0