Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C_398236253_10
          See other EPB41L2 GT Assays ›
          SNP ID:
          rs769106779
          Gene
          EPB41L2 SMLR1
          Gene Name
          erythrocyte membrane protein band 4.1 like 2
          small leucine rich protein 1
          Set Membership:
          -
          Chromosome Location:
          Chr.6: 130833470 - 130833470 on Build GRCh38
          Polymorphism:
          G/A, Transition substitution
          Context Sequence [VIC/FAM]:

          AGTGAGTTAAAAGTCTAAGCCATGG[G/A]CCATCTCCAAGGAATACCTTTGAAA

          Assay ID C_398236253_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 603237

          Literature Links:

          EPB41L2 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          EPB41L2 - erythrocyte membrane protein band 4.1 like 2
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001135554.1 Intron NP_001129026.1
          NM_001135555.3 Intron NP_001129027.1
          NM_001199388.2 Intron NP_001186317.1
          NM_001252660.1 Intron NP_001239589.1
          NM_001431.3 Intron NP_001422.1
          XM_006715362.2 Intron XP_006715425.1
          XM_011535524.1 Intron XP_011533826.1
          XM_011535525.1 Intron XP_011533827.1
          XM_011535527.1 Intron XP_011533829.1
          XM_011535528.1 Intron XP_011533830.1
          XM_011535529.1 Intron XP_011533831.1
          XM_011535530.1 Intron XP_011533832.1
          XM_011535534.1 Intron XP_011533836.1
          XM_011535536.1 Intron XP_011533838.1
          XM_017010349.1 Intron XP_016865838.1
          XM_017010350.1 Intron XP_016865839.1
          XM_017010351.1 Intron XP_016865840.1
          XM_017010352.1 Intron XP_016865841.1
          XM_017010353.1 Intron XP_016865842.1
          XM_017010354.1 Intron XP_016865843.1
          XM_017010355.1 Intron XP_016865844.1
          XM_017010356.1 Intron XP_016865845.1
          XM_017010357.1 Intron XP_016865846.1
          XM_017010358.1 Intron XP_016865847.1
          XM_017010359.1 Intron XP_016865848.1
          XM_017010360.1 Intron XP_016865849.1
          XM_017010361.1 Intron XP_016865850.1
          XM_017010362.1 Intron XP_016865851.1
          XM_017010363.1 Intron XP_016865852.1
          XM_017010364.1 Intron XP_016865853.1
          SMLR1 - small leucine rich protein 1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001195597.1 Intron NP_001182526.1

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cortical actin cytoskeleton organization
          actin binding
          structural molecule activity
          spectrin binding
          PH domain binding
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline