Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CATTCCGCTGCTGCTCGCCCCTCCT[C/T]CTGCAGGCGAAAGCAAGAAGATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120140 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COL2A1 - collagen type II alpha 1 chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM106C - transmembrane protein 106C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143841.1 | 224 | Missense Mutation | TCC,TTC | S,F 13 | NP_001137313.1 | |
NM_001143842.1 | 224 | Missense Mutation | TCC,TTC | S,F 13 | NP_001137314.1 | |
NM_001143843.1 | 224 | Missense Mutation | TCC,TTC | S,F 13 | NP_001137315.1 | |
NM_024056.3 | 224 | Missense Mutation | TCC,TTC | S,F 13 | NP_076961.1 |