Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CCCGGCCCCGGTTCCCACAGGACAC[A/G]CTAAGAAGCACAGGGAGCATTTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603924 MIM: 602873 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HABP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HABP2 - hyaluronan binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177660.2 | 2615 | UTR 3 | NP_001171131.1 | |||
NM_004132.4 | 2615 | UTR 3 | NP_004123.1 |
NRAP - nebulin related anchoring protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261463.1 | 2615 | Intron | NP_001248392.1 | |||
NM_001322945.1 | 2615 | Intron | NP_001309874.1 | |||
NM_006175.4 | 2615 | Intron | NP_006166.3 | |||
NM_198060.3 | 2615 | Intron | NP_932326.2 | |||
XM_005269864.2 | 2615 | Intron | XP_005269921.1 | |||
XM_005269865.2 | 2615 | Intron | XP_005269922.1 | |||
XM_005269867.2 | 2615 | Intron | XP_005269924.1 | |||
XM_006717870.2 | 2615 | Intron | XP_006717933.1 | |||
XM_011539832.2 | 2615 | Intron | XP_011538134.1 |
Set Membership: |
HapMap |