Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__63099243_10
          See other FLJ46906 GT Assays ›
          SNP ID:
          rs9495206
          Gene
          FLJ46906 NHSL1
          Gene Name
          uncharacterized LOC441172
          NHS like 1
          Set Membership:
          -
          Chromosome Location:
          Chr.6: 138688925 - 138688925 on Build GRCh38
          Polymorphism:
          A/T, Transversion substitution
          Context Sequence [VIC/FAM]:

          AAAGTAATTGTCTCGATCACTTACA[A/T]TGGTAGCAGTATTGTATTTTTAGTA

          Assay ID C__63099243_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          Literature Links:

          FLJ46906 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          A (0.20)
          (0.80)
          Caucasian - Not Available CEPH (CEU) - Not Available
          EAS
          A (0.38)
          (0.62)
          African American - Not Available YRI (Yoruba) - Not Available
          SAS
          A (0.12)
          (0.88)
          Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR
          A (0.19)
          (0.81)
          Japanese - Not Available JPT (Japanese) - Not Available
          EUR
          A (0.13)
          (0.87)
          AMR
          A (0.17)
          (0.83)
          FLJ46906 - uncharacterized LOC441172
          There are no transcripts associated with this gene.
          NHSL1 - NHS like 1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001144060.1 Intron NP_001137532.1
          NM_020464.1 Intron NP_065197.1
          XM_005267062.4 Intron XP_005267119.1
          XM_011535965.2 Intron XP_011534267.1
          XM_011535966.2 Intron XP_011534268.1
          XM_011535967.2 Intron XP_011534269.1
          XM_011535968.2 Intron XP_011534270.1
          XM_011535969.2 Intron XP_011534271.1
          XM_011535970.2 Intron XP_011534272.1
          XM_011535971.2 Intron XP_011534273.1
          XM_011535972.2 Intron XP_011534274.1
          XM_011535973.2 Intron XP_011534275.1
          XM_011535974.2 Intron XP_011534276.1
          XM_011535976.1 Intron XP_011534278.1
          XM_017011086.1 Intron XP_016866575.1
          XM_017011087.1 Intron XP_016866576.1
          XM_017011088.1 Intron XP_016866577.1
          XM_017011089.1 Intron XP_016866578.1

          Back To Top

          More Information



          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline