Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CATTCATGGTATGGGAACCTGTTCA[C/T]ATTTAAAATTACTTTGTGATCTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPHKAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPHKAP - SPHK1 interactor, AKAP domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142644.1 | 7027 | UTR 3 | NP_001136116.1 | |||
NM_030623.3 | 7027 | UTR 3 | NP_085126.2 | |||
XM_005246870.4 | 7027 | UTR 3 | XP_005246927.1 | |||
XM_006712777.3 | 7027 | UTR 3 | XP_006712840.1 | |||
XM_006712778.3 | 7027 | UTR 3 | XP_006712841.1 | |||
XM_011511925.2 | 7027 | UTR 3 | XP_011510227.1 |