Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CCCACAGAGCTTGCCCCTCAGTTCC[T/C]ACCTCCAAGGGGGAGGGTCCTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608682 MIM: 606774 MIM: 603560 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ADM2 - adrenomedullin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253845.1 | 1710 | UTR 3 | NP_001240774.1 | |||
XM_017028940.1 | 1710 | UTR 3 | XP_016884429.1 |
MIOX - myo-inositol oxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017584.5 | 1710 | Intron | NP_060054.4 | |||
XM_005261925.3 | 1710 | Intron | XP_005261982.1 | |||
XM_011530705.1 | 1710 | UTR 5 | XP_011529007.1 |
SBF1 - SET binding factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |