Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000526
          Assay Name: hsa-miR-223
          miRBase Accession Number: MI0000963
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 65367812 - 65367921 [+] on Build Rnor_6.0
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Species: Rat, Chicken, Dog, Fugu rubripes, Gorilla gorilla, Monodelphis domestica, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Saguinus labiatus, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus laevis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000526
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000067, mir-223

          Mature miRNA Sequence:

          UGUCAGUUUGUCAAAUACCCC

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-223-3p MIMAT0000892
          Chicken gga-miR-223 MIMAT0001140
          Dog cfa-miR-223 MIMAT0009852
          Fugu rubripes fru-miR-223 MIMAT0002894
          Gorilla gorilla ggo-miR-223 MIMAT0002590
          Monodelphis domestica mdo-miR-223-3p MIMAT0004135
          View More Rows
          Pan paniscus ppa-miR-223 MIMAT0002593
          Pan troglodytes ptr-miR-223 MIMAT0002589
          Pig ssc-miR-223 MIMAT0041607
          Pongo pygmaeus ppy-miR-223 MIMAT0002591
          Rhesus monkey mml-miR-223 MIMAT0002588
          Saguinus labiatus sla-miR-223 MIMAT0002592
          Taeniopygia guttata tgu-miR-223 MIMAT0014599
          Tetraodon nigroviridis tni-miR-223 MIMAT0002895
          Xenopus laevis xla-miR-223 MIMAT0011144
          Xenopus tropicalis xtr-miR-223 MIMAT0003634
          Zebrafish dre-miR-223 MIMAT0001290

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-223
          Stem-loop Accession # MI0000963
          Stem-loop Sequence
          UCUGGCCUUCUGCAGUGUUACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCUGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGUGGCUCAUGCUUAUCAG
          Chromosome Location Chr. X - 65367812 - 65367921 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000892
          miRBase ID: rno-miR-223-3p
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: Chr. X - 65367812 - 65367921 [+] on Build Rnor_6.0

          Chicken

          Stem-loop ID gga-mir-223
          Stem-loop Accession # MI0001208
          Stem-loop Sequence
          CUGCAGUGCAGCACUGCGUGUAUUUGACAAGCUGAGUUUGACACUCAGUCUGGCAGAGUGUCAGUUUGUCAAAUACCCCAAGUGAGGCACUUGCUGAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001140
          miRBase ID: gga-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-223
          Stem-loop Accession # MI0010347
          Stem-loop Sequence
          CCUGGCCUCCUGCAGUGCCACACUCCGUGUAUUUGACAAGCUGAGUUAGACACUCCAUGUGGAAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCACAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009852
          miRBase ID: cfa-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-223
          Stem-loop Accession # MI0003209
          Stem-loop Sequence
          CACUUAGUGUAUUUGACAAGCUGUUCUUGACACUCUUUAUACGCGAGUGUCAGUUUGUCAAAUACCCCAAGUGAGGUGUCACUUGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002894
          miRBase ID: fru-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-223
          Stem-loop Accession # MI0002899
          Stem-loop Sequence
          CCUGGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCACAUGCUUACCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002590
          miRBase ID: ggo-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-223
          Stem-loop Accession # MI0005324
          Stem-loop Sequence
          UCUGGCCCAGAUCCUUCAGUGCCACACUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCGUGUCGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGAGGCAUUUGCCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004135
          miRBase ID: mdo-miR-223-3p
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-223
          Stem-loop Accession # MI0002902
          Stem-loop Sequence
          CCUGGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCACAUGCUUACCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002593
          miRBase ID: ppa-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-223
          Stem-loop Accession # MI0002898
          Stem-loop Sequence
          CCUGGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCACAUGCUUACCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002589
          miRBase ID: ptr-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-223
          Stem-loop Accession # MI0033408
          Stem-loop Sequence
          CCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUAGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041607
          miRBase ID: ssc-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-223
          Stem-loop Accession # MI0002900
          Stem-loop Sequence
          CCUGGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUAGACACUCCGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCACAUGCUUACCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002591
          miRBase ID: ppy-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-223
          Stem-loop Accession # MI0002897
          Stem-loop Sequence
          CCCGGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCAUAUGCUUACCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002588
          miRBase ID: mml-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-223
          Stem-loop Accession # MI0002901
          Stem-loop Sequence
          CCUGGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGACACAUGCUUAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002592
          miRBase ID: sla-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-223
          Stem-loop Accession # MI0013827
          Stem-loop Sequence
          GUGUAUUUGACAAGCUGAGUCCGACACUCAGUGUGCAGAGUGUCAGUUUGUCAAAUACCCCAAGUGAGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014599
          miRBase ID: tgu-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-223
          Stem-loop Accession # MI0003210
          Stem-loop Sequence
          CAGGCCCUUCACUUAGUGUAUUUGACAAGCUGUGUUUGACACUCUGUAUCUGCGAGUGUCAGUUUGUCAAAUACCCCAAGUGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002895
          miRBase ID: tni-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-223
          Stem-loop Accession # MI0008382
          Stem-loop Sequence
          UGUGUGGCACUGAGUGUAUUUGACAAGCUGAGUCCGACACUCAGUGAGACAGAGUGUCAGUUUGUCAAAUACCCCAAGUGAGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011144
          miRBase ID: xla-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-223
          Stem-loop Accession # MI0004875
          Stem-loop Sequence
          AGUGUGGCACUGAGUGUAUUUGACAAGCUGAGUCCGACACUCAAUGAGACAGAGUGUCAGUUUGUCAAAUACCCCAAGUGAGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003634
          miRBase ID: xtr-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-223
          Stem-loop Accession # MI0001389
          Stem-loop Sequence
          CUCUCCUCCUGAUCUAGACUCUUCUCUUAGAGUAUUUGACAGACUGUGGUUGACACUCGAUCUAAAGGGGUGUCAGUUUGUCAAAUACCCCAAGAGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001290
          miRBase ID: dre-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCC
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Rodent Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Rodent MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Rodent MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465479_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10903
          Ambion® Pre-miR™ miRNA Precursor : PM10903
          mirVana® miRNA inhibitor : MH10903
          mirVana® miRNA mimic : MC10903
          TaqMan™ Advanced miRNA Assay : rno481007_mir
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          FGMIR : TaqManTM Array Rodent MicroRNA B Card v3
          MIRNA : TaqMan® Array Rodent MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v3.0
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool B v3.0


          miRBase Alias:

          Human: hsa-miR-223 (v9.2)
          Mouse: mmu-miR-223 (v9.2)
          Rat: rno-miR-223 (v18)
          Monodelphis domestica: mdo-miR-223 (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0