Search
Search

| Assay Name: | rno-miR-223-3p |
|---|---|
| miRBase Accession Number: | MI0000963 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUCAGUUUGUCAAAUACCCC |
| Species: | Rat, Chicken, Dog, Fugu rubripes, Gorilla gorilla, Monodelphis domestica, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Saguinus labiatus, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus laevis, Xenopus tropicalis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000067, mir-223 |
Mature miRNA Sequence: | UGUCAGUUUGUCAAAUACCCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-223-3p | MIMAT0000892 |
| Chicken | gga-miR-223 | MIMAT0001140 |
| Dog | cfa-miR-223 | MIMAT0009852 |
| Fugu rubripes | fru-miR-223 | MIMAT0002894 |
| Gorilla gorilla | ggo-miR-223 | MIMAT0002590 |
| Monodelphis domestica | mdo-miR-223-3p | MIMAT0004135 |
| View More Rows | ||
| Pan paniscus | ppa-miR-223 | MIMAT0002593 |
| Pan troglodytes | ptr-miR-223 | MIMAT0002589 |
| Pig | ssc-miR-223 | MIMAT0041607 |
| Pongo pygmaeus | ppy-miR-223 | MIMAT0002591 |
| Rhesus monkey | mml-miR-223 | MIMAT0002588 |
| Saguinus labiatus | sla-miR-223 | MIMAT0002592 |
| Taeniopygia guttata | tgu-miR-223 | MIMAT0014599 |
| Tetraodon nigroviridis | tni-miR-223 | MIMAT0002895 |
| Xenopus laevis | xla-miR-223 | MIMAT0011144 |
| Xenopus tropicalis | xtr-miR-223 | MIMAT0003634 |
| Zebrafish | dre-miR-223 | MIMAT0001290 |