Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002172
          Assay Name: hsa-let-7i*
          miRBase Accession Number: MI0000434
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 62603686 - 62603769 [+] on Build GRCh38
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Artibeus jamaicensis, Chrysemys picta, Columba livia, Dasypus novemcinctus, Goat, Guinea pig, Pig, Python bivittatus, Rabbit, Taeniopygia guttata
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002172
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUGCGCAAGCUACUGCCUUGCU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7i-3p MIMAT0004585
          Mouse mmu-let-7i-3p MIMAT0004520
          Rat rno-let-7i-3p MIMAT0004707
          Alligator mississippiensis ami-let-7i-3p MIMAT0038017
          Artibeus jamaicensis aja-let-7i MIMAT0031151
          Chrysemys picta cpi-let-7i-3p MIMAT0037611
          View More Rows
          Columba livia cli-let-7i-3p MIMAT0038390
          Dasypus novemcinctus dno-let-7i-3p MIMAT0047487
          Goat chi-let-7i-3p MIMAT0035895
          Guinea pig cpo-let-7i-3p MIMAT0046834
          Pig ssc-let-7i-3p MIMAT0037319
          Python bivittatus pbv-let-7i-3p MIMAT0038811
          Rabbit ocu-let-7i-3p MIMAT0048090
          Taeniopygia guttata tgu-let-7i-3p MIMAT0026974

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7i
          Stem-loop Accession # MI0000434
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location Chr. 12 - 62603686 - 62603769 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004585
          miRBase ID: hsa-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: Chr. 12 - 62603686 - 62603769 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7i
          Stem-loop Accession # MI0000138
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location Chr. 10 - 122985640 - 122985724 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004520
          miRBase ID: mmu-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: Chr. 10 - 122985640 - 122985724 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7i
          Stem-loop Accession # MI0000835
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location Chr. 7 - 66802731 - 66802815 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004707
          miRBase ID: rno-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: Chr. 7 - 66802731 - 66802815 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-let-7i
          Stem-loop Accession # MI0029651
          Stem-loop Sequence
          CGGUCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACACUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038017
          miRBase ID: ami-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Artibeus jamaicensis

          Stem-loop ID aja-let-7i
          Stem-loop Accession # MI0024373
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031151
          miRBase ID: aja-let-7i
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7i
          Stem-loop Accession # MI0029383
          Stem-loop Sequence
          CCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037611
          miRBase ID: cpi-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-let-7i
          Stem-loop Accession # MI0029892
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038390
          miRBase ID: cli-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7i
          Stem-loop Accession # MI0038894
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047487
          miRBase ID: dno-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7i
          Stem-loop Accession # MI0030586
          Stem-loop Sequence
          CCCGACACCAUGGCCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035895
          miRBase ID: chi-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7i
          Stem-loop Accession # MI0038550
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046834
          miRBase ID: cpo-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-let-7i
          Stem-loop Accession # MI0002447
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037319
          miRBase ID: ssc-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7i
          Stem-loop Accession # MI0030142
          Stem-loop Sequence
          ACCCUGGCUGAGGUAGUAGUUUGUGCUGUUCGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038811
          miRBase ID: pbv-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-let-7i
          Stem-loop Accession # MI0039214
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048090
          miRBase ID: ocu-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7i
          Stem-loop Accession # MI0013694
          Stem-loop Sequence
          GGGCCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026974
          miRBase ID: tgu-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool B v3.0
          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Cards:

          TaqMan® Human MicroRNA B Cards v3.0
          TaqMan® Rodent MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panels:

          TaqMan® OpenArray® Human MicroRNA Panel
          TaqMan® OpenArray® Rodent MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306160_pri
          TaqMan™ Pri-miRNA Assay : Rn03465226_pri
          TaqMan™ Pri-miRNA Assay : Hs03303039_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12783
          Ambion® Pre-miR™ miRNA Precursor : PM12783
          mirVana® miRNA inhibitor : MH12783
          mirVana® miRNA mimic : MC12783
          TaqMan™ Advanced miRNA Assay : rno480880_mir
          TaqMan™ Advanced miRNA Assay : 477862_mir
          TaqMan™ Advanced miRNA Assay : mmu481511_mir
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool B v3.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA B Cards v3.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          FGMIR : TaqManTM Human MicroRNA Array B v3
          FGMIR : TaqManTM Array Rodent MicroRNA B Card v3
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v3.0


          miRBase Alias:

          Human: hsa-let-7i* (v17)
          Mouse: mmu-let-7i* (v17)
          Rat: rno-let-7i* (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-snt5b:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline