Search
Search

| Assay Name: | hsa-miR-205-5p |
|---|---|
| miRBase Accession Number: | MI0000285 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCUUCAUUCCACCGGAGUCUG |
| Species: | Human, Mouse, Alligator mississippiensis, Anolis carolinensis, Astatotilapia burtoni, Ateles geoffroyi, Bovine, Chicken, Chrysemys picta, Columba livia, Cyprinus carpio, Dog, Fugu rubripes, Gorilla gorilla, Horse, Lagothrix lagotricha, Macaca nemestrina, Metriaclima zebra, Monodelphis domestica, Neolamprologus brichardi, Oreochromis niloticus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pundamilia nyererei, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Tetraodon nigroviridis, Xenopus laevis, Xenopus tropicalis, Zebrafish |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000058, mir-205 |
Mature miRNA Sequence: | UCCUUCAUUCCACCGGAGUCUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-205-5p | MIMAT0000266 |
| Mouse | mmu-miR-205-5p | MIMAT0000238 |
| Alligator mississippiensis | ami-miR-205a-5p | MIMAT0038228 |
| Anolis carolinensis | aca-miR-205a | MIMAT0021855 |
| Astatotilapia burtoni | abu-miR-205 | MIMAT0042062 |
| Ateles geoffroyi | age-miR-205 | MIMAT0002542 |
| View More Rows | ||
| Bovine | bta-miR-205 | MIMAT0003545 |
| Chicken | gga-miR-205a | MIMAT0001184 |
| Chrysemys picta | cpi-miR-205a-5p | MIMAT0037830 |
| Columba livia | cli-miR-205a-5p | MIMAT0038584 |
| Cyprinus carpio | ccr-miR-205 | MIMAT0026263 |
| Dog | cfa-miR-205 | MIMAT0009845 |
| Fugu rubripes | fru-miR-205 | MIMAT0002965 |
| Gorilla gorilla | ggo-miR-205 | MIMAT0002541 |
| Horse | eca-miR-205 | MIMAT0012962 |
| Lagothrix lagotricha | lla-miR-205 | MIMAT0002545 |
| Macaca nemestrina | mne-miR-205 | MIMAT0002546 |
| Metriaclima zebra | mze-miR-205 | MIMAT0042296 |
| Monodelphis domestica | mdo-miR-205a | MIMAT0012749 |
| Neolamprologus brichardi | nbr-miR-205 | MIMAT0042484 |
| Oreochromis niloticus | oni-miR-205 | MIMAT0042668 |
| Pan paniscus | ppa-miR-205 | MIMAT0002543 |
| Pan troglodytes | ptr-miR-205 | MIMAT0002544 |
| Pig | ssc-miR-205 | MIMAT0002146 |
| Pongo pygmaeus | ppy-miR-205 | MIMAT0015798 |
| Pundamilia nyererei | pny-miR-205 | MIMAT0042873 |
| Python bivittatus | pbv-miR-205a-5p | MIMAT0039009 |
| Rabbit | ocu-miR-205-5p | MIMAT0048325 |
| Rhesus monkey | mml-miR-205 | MIMAT0006236 |
| Salmo salar | ssa-miR-205b-5p | MIMAT0032495 |
| Tetraodon nigroviridis | tni-miR-205 | MIMAT0002966 |
| Xenopus laevis | xla-miR-205 | MIMAT0011142 |
| Xenopus tropicalis | xtr-miR-205a | MIMAT0003697 |
| Zebrafish | dre-miR-205-5p | MIMAT0001280 |