Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12670
          Assay Name: hsa-miR-340-5p
          miRBase Accession Number: MI0000802
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 180015303 - 180015397 [-] on Build GRCh38
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Species: Human, Mouse, Rat, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12670

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000191, mir-340

          Mature miRNA Sequence:

          UUAUAAAGCAAUGAGACUGAUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-340-5p MIMAT0004692
          Mouse mmu-miR-340-5p MIMAT0004651
          Rat rno-miR-340-5p MIMAT0004650
          Callithrix jacchus cja-miR-340 MIMAT0049315
          Cricetulus griseus cgr-miR-340-5p MIMAT0023920
          Dasypus novemcinctus dno-miR-340-5p MIMAT0047823
          View More Rows
          Daubentonia madagascariensis dma-miR-340 MIMAT0049244
          Dog cfa-miR-340 MIMAT0009892
          Goat chi-miR-340-5p MIMAT0036155
          Guinea pig cpo-miR-340-5p MIMAT0047178
          Horse eca-miR-340-5p MIMAT0013066
          Microcebus murinus mmr-miR-340 MIMAT0049159
          Nomascus leucogenys nle-miR-340 MIMAT0049343
          Otolemur garnettii oga-miR-340 MIMAT0049696
          Pan paniscus ppa-miR-340 MIMAT0049106
          Pan troglodytes ptr-miR-340 MIMAT0008109
          Papio hamadryas pha-miR-340 MIMAT0049556
          Pig ssc-miR-340 MIMAT0013891
          Pongo pygmaeus ppy-miR-340 MIMAT0015843
          Pteropus alecto pal-miR-340-5p MIMAT0040191
          Rabbit ocu-miR-340-5p MIMAT0048386
          Rhesus monkey mml-miR-340-5p MIMAT0006281

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-340
          Stem-loop Accession # MI0000802
          Stem-loop Sequence
          UUGUACCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUA
          Chromosome Location Chr. 5 - 180015303 - 180015397 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004692
          miRBase ID: hsa-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: Chr. 5 - 180015303 - 180015397 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-340
          Stem-loop Accession # MI0000623
          Stem-loop Sequence
          CAAUUGUACUUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUA
          Chromosome Location Chr. 11 - 50069702 - 50069799 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004651
          miRBase ID: mmu-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: Chr. 11 - 50069702 - 50069799 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-340-1
          Stem-loop Accession # MI0000622
          Stem-loop Sequence
          CACUUGUACUCGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUGUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUA
          Chromosome Location Chr. 10 - 35558983 - 35559080 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004650
          miRBase ID: rno-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: Chr. 10 - 35571755 - 35571852 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-340-2
          Stem-loop Accession # MI0031764
          Stem-loop Sequence
          CACUUGUACUCGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUGUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUA
          Chromosome Location Chr. 10 - 35571755 - 35571852 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004650
          miRBase ID: rno-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: Chr. 10 - 35571755 - 35571852 [+] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-340
          Stem-loop Accession # MI0040051
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUUGUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049315
          miRBase ID: cja-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-340
          Stem-loop Accession # MI0020501
          Stem-loop Sequence
          UUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023920
          miRBase ID: cgr-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-340
          Stem-loop Accession # MI0039076
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUAGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047823
          miRBase ID: dno-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-340
          Stem-loop Accession # MI0039972
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049244
          miRBase ID: dma-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-340
          Stem-loop Accession # MI0010391
          Stem-loop Sequence
          UUGUACCUGAUGCGAUUAUAAAGCAAUGAGACUGAUUGUCAUGUGUCGGUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCGUGCCUGGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009892
          miRBase ID: cfa-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-340
          Stem-loop Accession # MI0030751
          Stem-loop Sequence
          GUUUGUACCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUGUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUAUACCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036155
          miRBase ID: chi-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-340
          Stem-loop Accession # MI0038737
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUUUGUCUUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047178
          miRBase ID: cpo-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-340
          Stem-loop Accession # MI0012811
          Stem-loop Sequence
          GGUUAUAAAGCAAUGAGACUGAUUGUCAUGUGUCGCUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013066
          miRBase ID: eca-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-340
          Stem-loop Accession # MI0039875
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUUGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049159
          miRBase ID: mmr-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-340
          Stem-loop Accession # MI0040079
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049343
          miRBase ID: nle-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-340
          Stem-loop Accession # MI0040480
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCAUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049696
          miRBase ID: oga-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-340
          Stem-loop Accession # MI0039818
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049106
          miRBase ID: ppa-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-340
          Stem-loop Accession # MI0008628
          Stem-loop Sequence
          UUGUACCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008109
          miRBase ID: ptr-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-340
          Stem-loop Accession # MI0040326
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUUGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049556
          miRBase ID: pha-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-340-1
          Stem-loop Accession # MI0013110
          Stem-loop Sequence
          UACCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013891
          miRBase ID: ssc-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Stem-loop ID ssc-mir-340-2
          Stem-loop Accession # MI0022145
          Stem-loop Sequence
          CCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013891
          miRBase ID: ssc-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-340
          Stem-loop Accession # MI0014904
          Stem-loop Sequence
          UUGUAUCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUCGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015843
          miRBase ID: ppy-miR-340
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-340
          Stem-loop Accession # MI0032542
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUGUGUUGUGUGUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040191
          miRBase ID: pal-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-340
          Stem-loop Accession # MI0039373
          Stem-loop Sequence
          UUAUAAAGCAAUGAGACUGAUUGUCAUAUGUUGUUCAUGGGAUCCGUCUCAGUUACUUUAUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048386
          miRBase ID: ocu-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-340
          Stem-loop Accession # MI0007703
          Stem-loop Sequence
          UUGUACCUGGUGUGAUUAUAAAGCAAUGAGACUGAUUGUCAUAUGUUGUUUGUGGGAUCCGUCUCAGUUACUUUAUAGCCAUACCUGGUAUCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006281
          miRBase ID: mml-miR-340-5p
          Mature miRNA Sequence: UUAUAAAGCAAUGAGACUGAUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306995_pri
          TaqMan™ Pri-miRNA Assay : Hs03303572_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12670
          Ambion® Pre-miR™ miRNA Precursor : PM12670
          TaqMan™ MicroRNA Assay : 002258
          mirVana® miRNA inhibitor : MH12670
          TaqMan™ Advanced miRNA Assay : 478042_mir
          TaqMan™ Advanced miRNA Assay : rno481072_mir
          TaqMan™ Advanced miRNA Assay : mmu481072_mir


          miRBase Alias:

          Human: hsa-miR-340 (v17)
          Macaca mulatta: mml-miR-340 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline