Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12921
          Assay Name: hsa-miR-214-5p
          miRBase Accession Number: MI0000290
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 172138798 - 172138907 [-] on Build GRCh38
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Species: Human, Mouse, Alligator mississippiensis, Anolis carolinensis, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Monodelphis domestica, Pig, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Taeniopygia guttata
          Product Type: mirVana® miRNA mimic
          Assay ID MC12921

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000062, mir-214

          Mature miRNA Sequence:

          UGCCUGUCUACACUUGCUGUGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-214-5p MIMAT0004564
          Mouse mmu-miR-214-5p MIMAT0004664
          Alligator mississippiensis ami-miR-214-5p MIMAT0038238
          Anolis carolinensis aca-miR-214-5p MIMAT0021871
          Columba livia cli-miR-214-5p MIMAT0038592
          Cricetulus griseus cgr-miR-214-5p MIMAT0023851
          View More Rows
          Dasypus novemcinctus dno-miR-214-5p MIMAT0047758
          Goat chi-miR-214-5p MIMAT0036058
          Guinea pig cpo-miR-214-5p MIMAT0047115
          Monodelphis domestica mdo-miR-214-5p MIMAT0026679
          Pig ssc-miR-214-5p MIMAT0037318
          Pteropus alecto pal-miR-214-5p MIMAT0040067
          Python bivittatus pbv-miR-214-5p MIMAT0039018
          Rabbit ocu-miR-214-5p MIMAT0048337
          Rhesus monkey mml-miR-214-5p MIMAT0026588
          Salmo salar ssa-miR-214-5p MIMAT0032514
          Taeniopygia guttata tgu-miR-214-5p MIMAT0027021

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-214
          Stem-loop Accession # MI0000290
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location Chr. 1 - 172138798 - 172138907 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004564
          miRBase ID: hsa-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: Chr. 1 - 172138798 - 172138907 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-214
          Stem-loop Accession # MI0000698
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location Chr. 1 - 162223368 - 162223477 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004664
          miRBase ID: mmu-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: Chr. 1 - 162223368 - 162223477 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-214
          Stem-loop Accession # MI0029797
          Stem-loop Sequence
          CGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCACUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038238
          miRBase ID: ami-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-214
          Stem-loop Accession # MI0018817
          Stem-loop Sequence
          CAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021871
          miRBase ID: aca-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-214
          Stem-loop Accession # MI0030015
          Stem-loop Sequence
          GGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCGCAUGACAACCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038592
          miRBase ID: cli-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-214
          Stem-loop Accession # MI0020460
          Stem-loop Sequence
          GUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023851
          miRBase ID: cgr-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-214
          Stem-loop Accession # MI0039043
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047758
          miRBase ID: dno-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-214
          Stem-loop Accession # MI0030689
          Stem-loop Sequence
          GGCCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036058
          miRBase ID: chi-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-214
          Stem-loop Accession # MI0038705
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047115
          miRBase ID: cpo-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-214
          Stem-loop Accession # MI0005319
          Stem-loop Sequence
          GGGCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026679
          miRBase ID: mdo-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-214
          Stem-loop Accession # MI0002441
          Stem-loop Sequence
          GGCCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037318
          miRBase ID: ssc-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-214
          Stem-loop Accession # MI0032476
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040067
          miRBase ID: pal-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-214
          Stem-loop Accession # MI0030287
          Stem-loop Sequence
          UGGAUAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAAUCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039018
          miRBase ID: pbv-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-214
          Stem-loop Accession # MI0039348
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048337
          miRBase ID: ocu-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-214
          Stem-loop Accession # MI0002858
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026588
          miRBase ID: mml-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-214-1
          Stem-loop Accession # MI0026611
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACAUCCUGGAACCUGUACAGCAGGCACAGACAGGCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032514
          miRBase ID: ssa-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-214-2
          Stem-loop Accession # MI0026612
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGACCUUCCUAUCCUGUACAGCAGGCACAGACAGGCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032514
          miRBase ID: ssa-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-214-3
          Stem-loop Accession # MI0026613
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGAACUUCCUGGAACCUGUACAGCAGGCACAGACAGGCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032514
          miRBase ID: ssa-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-214
          Stem-loop Accession # MI0013833
          Stem-loop Sequence
          GACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCGCAUGACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0027021
          miRBase ID: tgu-miR-214-5p
          Mature miRNA Sequence: UGCCUGUCUACACUUGCUGUGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302971_pri
          TaqMan™ Pri-miRNA Assay : Mm03307122_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12921
          Ambion® Pre-miR™ miRNA Precursor : PM12921
          TaqMan™ MicroRNA Assay : 002293
          mirVana® miRNA inhibitor : MH12921
          TaqMan™ Advanced miRNA Assay : 478768_mir
          TaqMan™ Advanced miRNA Assay : mmu478768_mir


          miRBase Alias:

          Human: hsa-miR-214* (v17)
          Mouse: mmu-miR-214* (v17)
          Aeromonas caviae Ae398: aca-miR-214* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline