Search
Search

| Assay Name: | mmu-miR-1264-3p |
|---|---|
| miRBase Accession Number: | MI0004130 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAAUCUUAUUUGAGCACCUGU |
| Species: | Mouse |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | CAAAUCUUAUUUGAGCACCUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1264-3p | MIMAT0014803 |