Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC25489
          Assay Name: hsa-miR-139-3p
          miRBase Accession Number: MI0000261
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 72615063 - 72615130 [-] on Build GRCh38
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Species: Human, Cricetulus griseus, Dasypus novemcinctus, Eptesicus fuscus, Guinea pig, Pig, Pteropus alecto, Rabbit
          Product Type: mirVana® miRNA mimic
          Assay ID MC25489

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000117, mir-139

          Mature miRNA Sequence:

          UGGAGACGCGGCCCUGUUGGAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-139-3p MIMAT0004552
          Cricetulus griseus cgr-miR-139-3p MIMAT0023766
          Dasypus novemcinctus dno-miR-139-3p MIMAT0047652
          Eptesicus fuscus efu-miR-139 MIMAT0035163
          Guinea pig cpo-miR-139-3p MIMAT0047003
          Pig ssc-miR-139-3p MIMAT0022921
          View More Rows
          Pteropus alecto pal-miR-139-3p MIMAT0039926
          Rabbit ocu-miR-139-3p MIMAT0048240

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-139
          Stem-loop Accession # MI0000261
          Stem-loop Sequence
          GUGUAUUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAAC
          Chromosome Location Chr. 11 - 72615063 - 72615130 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004552
          miRBase ID: hsa-miR-139-3p
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location: Chr. 11 - 72615063 - 72615130 [-] on Build GRCh38

          Cricetulus griseus

          Stem-loop ID cgr-mir-139
          Stem-loop Accession # MI0020401
          Stem-loop Sequence
          CAGGUGUAUUCUACAGUGCACGUGUCUCCAGUGUGGCUCAGAGGCUGGAGACGCGGCCCUGUUGGAGUAACAACUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023766
          miRBase ID: cgr-miR-139-3p
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-139
          Stem-loop Accession # MI0038988
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047652
          miRBase ID: dno-miR-139-3p
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-139
          Stem-loop Accession # MI0028870
          Stem-loop Sequence
          UCAGGUGUAUUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAACAACUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035163
          miRBase ID: efu-miR-139
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-139
          Stem-loop Accession # MI0038646
          Stem-loop Sequence
          GUGUAUUCUACAGUGCACGUGUCUCCAGCGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047003
          miRBase ID: cpo-miR-139-3p
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-139
          Stem-loop Accession # MI0002453
          Stem-loop Sequence
          GUGUAUUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022921
          miRBase ID: ssc-miR-139-3p
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-139
          Stem-loop Accession # MI0032403
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUCAGAGGCUGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039926
          miRBase ID: pal-miR-139-3p
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-139
          Stem-loop Accession # MI0039297
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUUGGAGGCUGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048240
          miRBase ID: ocu-miR-139-3p
          Mature miRNA Sequence: UGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302845_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM25489
          Ambion® Pre-miR™ miRNA Precursor : PM25489
          mirVana® miRNA inhibitor : MH25489
          TaqMan™ Advanced miRNA Assay : 477906_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline