Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10332
          Assay Name: ath-miR159a
          miRBase Accession Number: MI0000189
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 27713233 - 27713416 [-] on Build TAIR10.1
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Species: Arabidopsis, Amborella trichopoda, Arabidopsis lyrata, Arachis hypogaea, Asparagus officinalis, Brassica napus, Brassica rapa, Carica papaya, Citrus sinensis, Cucumis melo, Fragaria vesca, Glycine max, Helianthus tuberosus, Hevea brasiliensis, Malus domestica, Manihot esculenta, Medicago truncatula, Nicotiana tabacum, Phaseolus vulgaris, Populus trichocarpa, Prunus persica, Ricinus communis, Solanum lycopersicum, Vitis vinifera, Vriesea carinata
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10332

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000010, MIR159

          Mature miRNA Sequence:

          UUUGGAUUGAAGGGAGCUCUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Arabidopsis ath-miR159a MIMAT0000177
          Amborella trichopoda atr-miR159 MIMAT0033911
          Arabidopsis lyrata aly-miR159a-3p MIMAT0017424
          Arachis hypogaea ahy-miR159 MIMAT0016321
          Asparagus officinalis aof-miR159 MIMAT0049768
          Brassica napus bna-miR159 MIMAT0005635
          View More Rows
          Brassica rapa bra-miR159a MIMAT0010153
          Carica papaya cpa-miR159a MIMAT0031780
          Citrus sinensis csi-miR159a-3p MIMAT0018450
          Cucumis melo cme-miR159a MIMAT0026160
          Fragaria vesca fve-miR159a-3p MIMAT0044488
          Glycine max gma-miR159a-3p MIMAT0001675
          Glycine max gma-miR159e-3p MIMAT0021641
          Helianthus tuberosus htu-miR159a MIMAT0025497
          Hevea brasiliensis hbr-miR159a MIMAT0025283
          Malus domestica mdm-miR159d MIMAT0043539
          Malus domestica mdm-miR159e MIMAT0043540
          Malus domestica mdm-miR159f MIMAT0043541
          Manihot esculenta mes-miR159a-3p MIMAT0024410
          Manihot esculenta mes-miR159b MIMAT0029176
          Medicago truncatula mtr-miR159a MIMAT0011166
          Nicotiana tabacum nta-miR159 MIMAT0024639
          Phaseolus vulgaris pvu-miR159a.1 MIMAT0015301
          Populus trichocarpa ptc-miR159a MIMAT0001901
          Populus trichocarpa ptc-miR159b MIMAT0001902
          Prunus persica ppe-miR159 MIMAT0031437
          Ricinus communis rco-miR159 MIMAT0014152
          Solanum lycopersicum sly-miR159 MIMAT0009141
          Vitis vinifera vvi-miR159c MIMAT0005650
          Vriesea carinata vca-miR159-3p MIMAT0040991

          Stem-loop Details



          Arabidopsis

          Stem-loop ID ath-MIR159a
          Stem-loop Accession # MI0000189
          Stem-loop Sequence
          GUAGAGCUCCUUAAAGUUCAAACAUGAGUUGAGCAGGGUAAAGAAAAGCUGCUAAGCUAUGGAUCCCAUAAGCCCUAAUCCUUGUAAAGUAAAAAAGGAUUUGGUUAUAUGGAUUGCAUAUCUCAGGAGCUUUAACUUGCCCUUUAAUGGCUUUUACUCUUCUUUGGAUUGAAGGGAGCUCUAC
          Chromosome Location Chr. 1 - 27713233 - 27713416 [-] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0000177
          miRBase ID: ath-miR159a
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: Chr. 1 - 27713233 - 27713416 [-] on Build TAIR10.1

          Amborella trichopoda

          Stem-loop ID atr-MIR159
          Stem-loop Accession # MI0027509
          Stem-loop Sequence
          UUCUAUGAGUUGUUUAGAUUGUUGGGUUGUGUGUAUUUUUGUAGGGUUGUUGACAUGGAGCUCCUUGAGGUCCAAUCCAGGUUCUUACUUAGGUAAAAGCAGCUGCUAGUUCAUGGAUCCCACUGUGUUAUUUCCAAAUAUAAGAGAAUGGAGAUAUAAUGGUGGUUUGCAUGACCAAGGAGCUGCAUUCUUGCCUUGUGAGAUUCCUUGUUUGGAUUGAAGGGAGCUCUAUGUGUUCUCCCCUUCCAAGCUUAUUUUCUCUCUCAUUUAUUUAGCCCUUGAUCUUUUCAUUUUUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0033911
          miRBase ID: atr-miR159
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Arabidopsis lyrata

          Stem-loop ID aly-MIR159a
          Stem-loop Accession # MI0014515
          Stem-loop Sequence
          GAGAAAAAAAAAGAUGGAAGUAGAGCUCCUUGAAGUUCAAACGAGAGUUGAGCAGGGUAAAGAAAAGCUGCUAAGCUAUGGAUCCCAUAAGCCCUAAUCCUUAUAAAGAAAAAAAAGGAUUUGGUUAUAUGGCUUGCAUAUCUCAGGAGCUUUAACUUGCCCUUUAAUGGCUUUUACUCUUCUUUGGAUUGAAGGGAGCUCUACAUCUUCUUUCACCUUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017424
          miRBase ID: aly-miR159a-3p
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Arachis hypogaea

          Stem-loop ID ahy-MIR159
          Stem-loop Accession # MI0015335
          Stem-loop Sequence
          AGUGGAGCUCCUUGAAGUCCAACAGAGGGUCUUGGUGGGUAGAUUGAGCUGCUAAGCUAUGGAUCCCACAGUACUACCCAUAUAUACCCCAUUAUGCAGUGGUAGUUCUGCGGAUUCCAUAUCCUGGGAGCUUCAUUACCUUUCCACAGAUCCUUAUUUGGAUUGAAGGGAGCUCUACAUAGCAUCCUCUUUCUAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016321
          miRBase ID: ahy-miR159
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Asparagus officinalis

          Stem-loop ID aof-MIR159
          Stem-loop Accession # MI0040562
          Stem-loop Sequence
          CAUAAAAUUGUAGGUUUUAAGAGUGGAGCUCCUUGAAGUCCAAAUAUGGGUAUUGGAGGGGUGGAUCGAGCUGCUUUGUCAUGGAUUCCACUGCCCUAUUCCUAUAUGUUAUUUUCAUCUUGAGAGAAAAAAAAAGGAGGGGGGGAGAAAGAAAAAGAUCAUGUUCAUGGGAUAGGUUUGUGGCUUGCAUGACUCAGGAGCUUCAUUAACCUCUUCUUACCCUUCUUUGGAUUGAAGGGAGCUCUACACGUUAAUCUUUCUAAUUUUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049768
          miRBase ID: aof-miR159
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Brassica napus

          Stem-loop ID bna-MIR159
          Stem-loop Accession # MI0006480
          Stem-loop Sequence
          GUAGGGCUCCUUAUAGUUCAAACGAGAGUUUAGCAGGGUAAAGAAAAGCUGCUAAGCUAUGAAUCCCAUAAGCCCUAAUCCUUAUAUAUAAAAAGGAUUUGGUUAUAUGGCUUACAUAUCUCAGGAGCUUUAACUUGCCCUUUAAUUGCUUUUACUCUUCUUUGGAUUGAAGGGAGCUCUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005635
          miRBase ID: bna-miR159
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Brassica rapa

          Stem-loop ID bra-MIR159a
          Stem-loop Accession # MI0010649
          Stem-loop Sequence
          AAGUAGGGCUCCUUAUAGUUCAAACGAGAGUUUAGCAGGGUAAAGAAAAGCUGCUAAGCUAUGAAUCCCAUAAGCCCUAAUCCUUAUAUAUAAAAAGGAUUUGGUUAUAUGGCUUACAUAUCUCAGGAGCUUUAACUUGCCCUUUAAUUGCUUUUACUCUUCUUUGGAUUGAAGGGAGCUCUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010153
          miRBase ID: bra-miR159a
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Carica papaya

          Stem-loop ID cpa-MIR159a
          Stem-loop Accession # MI0026334
          Stem-loop Sequence
          GAGCUCCUUGAAGUCCAAUAGAGGAUUUUGCAGGGUUGAUUGAGCUGCCGAGCUAUGGAUUCCACAGCCCUACCCGAAGUUAUACUUUUUCUGGGUAGGCUUGCGGUUUGCAUAUCUCAGGAGCUGCAUUACCCUAAGCUUAAUCCUUGUUUGGAUUGAAGGGAGCUCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031780
          miRBase ID: cpa-miR159a
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Citrus sinensis

          Stem-loop ID csi-MIR159a
          Stem-loop Accession # MI0016693
          Stem-loop Sequence
          GGGUUAAGGAGUGGAGCUCCUUGAAGUCCAAAAGAGGUUCUAGCAAGGGUAAAUUGAGCUGCUGAGCUAUGGAUCCCACAGUCCUAUGUAUCAGAUUUUAGGGUUUGUUUCUGAUGAUGGUGAUGAUAGGUUUGUGGCUUGCAUAUUUCUGGAGCUUCAUUACCCCUUAAGUUUUACCCUUUUUUGGAUUGAAGGGAGCUCUACCUCUCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018450
          miRBase ID: csi-miR159a-3p
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Cucumis melo

          Stem-loop ID cme-MIR159a
          Stem-loop Accession # MI0023264
          Stem-loop Sequence
          GAGGCAGAGCUCCUUGAAGUCCAAUAGAGGUUUAUGCCAGGUUGAUUGAGCUGCUAAGCUAUGGAUCCCACAGCCCCUAUUGCUAUAUCCAUCAUGUUGUUGUAUAAUAUCUUUAAAGCUGAUAGGCCUGUGGCUUGCAUAUCUCAGGAGCUUCAUCACCUGUUUCUGUUAGAUCCUUGUUUGGAUUGAAGGGAGCUCUACAUCUUCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026160
          miRBase ID: cme-miR159a
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Fragaria vesca

          Stem-loop ID fve-MIR159a
          Stem-loop Accession # MI0036360
          Stem-loop Sequence
          AGCUAGGGUUAUGGAGUGGAGCUCCUUGAAGUCCAAUAGAAGAUGUUGCUAGGUUGAUUGAGCUGCUGAGCUAUGAAUUCCACACUUCUAUCUCUGUUUUUUGAUAAAGAUGAUAGGCUUGUGGCUUGCAUACCUCAGGAGCUGCAUUACCUGAGUUGCAUCAUGUUUUGGAUUGAAGGGAGCUCUACACCUUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044488
          miRBase ID: fve-miR159a-3p
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Glycine max

          Stem-loop ID gma-MIR159a
          Stem-loop Accession # MI0001773
          Stem-loop Sequence
          AAUUAAAGGGGAUUAUGAAGUGGAGCUCCUUGAAGUCCAAUUGAGGAUCUUACUGGGUGAAUUGAGCUGCUUAGCUAUGGAUCCCACAGUUCUACCCAUCAAUAAGUGCUUUUGUGGUAGUCUUGUGGCUUCCAUAUCUGGGGAGCUUCAUUUGCCUUUAUAGUAUUAACCUUCUUUGGAUUGAAGGGAGCUCUACACCCUUCUCUUCUUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001675
          miRBase ID: gma-miR159a-3p
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Stem-loop ID gma-MIR159e
          Stem-loop Accession # MI0018657
          Stem-loop Sequence
          CAAAGGGGGUUAUGGAGUGGAGCUCCUUGAAGUCCAAUUGAGGAUCUUACUGGGUGGAUUGAGCUGCUUAGCUAUGGAUCCCACAGUUCUACCCAUCAUUAAGUGCUUUUGUGGUAGUCUUGUGGCUUCCAUAUCUGGGGAGCUUCAUUUGCCUUUAUAGUAUUAUCCUUCUUUGGAUUGAAGGGAGCUCUACACCCUUCUCCCCUUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021641
          miRBase ID: gma-miR159e-3p
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Helianthus tuberosus

          Stem-loop ID htu-MIR159a
          Stem-loop Accession # MI0022237
          Stem-loop Sequence
          GUAGAGCUCCUUAAAGUCCAAUAGAAGAUCUGAGUGAGAACAUAGAGCUGCUAGUUUAUGGAUCCCUCAAGUCCUAUCUUUGUUGAUGUAUAGGCUUUGUGGCUUGCAUACACUUGGAGCUUUAUUACCCUCCUUAUGAUCCUUUUUUGGAUUGAAGGGAGCUCUACACCAUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025497
          miRBase ID: htu-miR159a
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Hevea brasiliensis

          Stem-loop ID hbr-MIR159a
          Stem-loop Accession # MI0022053
          Stem-loop Sequence
          UUGAAUACUGAUUUAGGGUUAAGAAGUGGAGCUCCUUGAAGUCCAAUAGAGGUUCUUGCUGGGUAGAUUGAGCUGCUGAGCUAUGGAUCCCUCAGCCCUAUCCCAUCAUAAAGAGUAGUCCUCUUGGAUAGGCUUGUGGCUUGCAUAUCUCAGGAGCUUCAUUACCUCAUAUGUUAGAUCCUUGUUUGGAUUGAAGGGAGCUCUACAUCUAAGUCUUUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025283
          miRBase ID: hbr-miR159a
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Malus domestica

          Stem-loop ID mdm-MIR159d
          Stem-loop Accession # MI0035594
          Stem-loop Sequence
          GGUUAAGGAUUGGAGCUCCUUGAAGUCCAAAAGAAGGUUAUGCUGGGUGGAUAGAGCUGUUGAGCUAUGGAUCCCACAGUACUAUUUAUCCUAUGUUAGAUCAGAUCGGAUCGACCGAAGGGAUGAUAGGGUUGUGGCUUGCAUAUCUCAGGAGCUGCAUUACCUGUACGCUGCCUCUUUUUUUGGAUUGAAGGGAGCUCUACACCUCUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0043539
          miRBase ID: mdm-miR159d
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Stem-loop ID mdm-MIR159e
          Stem-loop Accession # MI0035595
          Stem-loop Sequence
          AUAUAUAUGUAUGUGGGUAGGGUUAAGGAGUGGAGCUCCUUGAAGUCCAAAAGAAGGUGAUGCUGGGUGGAUAGAGCUGCUGAGCUAUGGAUCCCAGAGUCCUAUAUAUCCUAUAUCGCAGAUCAGAUCAGAUGCGAUCAGAAGGAUGAUAGGCUUGUGGCUUGCAUAUCUCAGGAGCUGCAUUACCUGUACGCUGCCUCUUUUUUUGGAUUGAAGGGAGCUCUACACCUCUCUCUCUGUCUCUCUCUCUCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0043540
          miRBase ID: mdm-miR159e
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Stem-loop ID mdm-MIR159f
          Stem-loop Accession # MI0035596
          Stem-loop Sequence
          AUGUGGGUAGGGUUAAGGAGUGGAGCUCCUUGAAGUCCAAAAGAAGGUGAUGCUGGGUGGAUAGAGCUGCUGAGCUAUGGAUCCCAGAGUCCUAUAUAUCCUAUAUCGCAGAUCAGAUCAGAUGCGAUCAGAAGGAUGAUAGGCUUGUGGCUUGCAUAUCUCAGGAGCUGCAUUACCUGUACGCUGCCUCUUUUUUUGGAUUGAAGGGAGCUCUACACCUCUCUCUCUGUCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0043541
          miRBase ID: mdm-miR159f
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Manihot esculenta

          Stem-loop ID mes-MIR159a
          Stem-loop Accession # MI0020959
          Stem-loop Sequence
          UGGAGCUCCUUGAAGUCCAAUAGAGGAUCUUGCUGGGUAGAUUGAGCUGCUGAGCUAUGGAUCCCACAGCCCUAUCUCAUAAUAAAGACUUUAGUCUUCUUGGAUAGGCUUGUGGCUUGCAUAUCUCAGGAGCUUCAUUACCUCUUAUGUUAGAUCCUUGUUUGGAUUGAAGGGAGCUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024410
          miRBase ID: mes-miR159a-3p
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Stem-loop ID mes-MIR159b
          Stem-loop Accession # MI0024218
          Stem-loop Sequence
          AGGGUUAAGAAGUGGAGCUCCUUAAAGUCCAAUAGAGGAUCUUGCAGGGUAGAUUGAGCUGCUGAGCUAUGGAUACCACAGCCCUAUCCCAUAAUAAAAGGGAGUUCUUUUUUUCAAGUCUUCUUGGAUAGGCUUGUGGCUUGCAUAUCUCUGGAGCUUCAUUACCUUAUUAUGUUAGAUCCUUGUUUGGAUUGAAGGGAGCUCUACAUCUAAAUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029176
          miRBase ID: mes-miR159b
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Medicago truncatula

          Stem-loop ID mtr-MIR159a
          Stem-loop Accession # MI0010696
          Stem-loop Sequence
          GGGGUGGAGCUUCCUUUAGUCCAAAUAUGGAUCUUGCUAUGUUGAUAGAGCUGCUUAGCUAUGGGUCCCUCAACUCUACCCAUCUUGUUCUUUGUGGUAGUUUUGUGGCUUCCAUAUCUAGGGAGCCUUAUCACCUUUAGUUUAAUCUUUCUUUGGAUUGAAGGGAGCUCUACAUCUUGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011166
          miRBase ID: mtr-miR159a
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Nicotiana tabacum

          Stem-loop ID nta-MIR159
          Stem-loop Accession # MI0021329
          Stem-loop Sequence
          UUUGAGAGUGGAGCUCCUUGAAGUCCAACAGAGGAUCUAACAGGUAAGAUUGAGCUGCUGACCUAUGGAUUCCUCAGCCCUAUCUAUUAUUUGAUUGGAUAGGUUUGUGGGUUGCAUAUGUCAGGAGCUUCAUUGCCCUAAGUUGGAUCCCUUUUUGGAUUGAAGGGAGCUCUACAUUCAUUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024639
          miRBase ID: nta-miR159
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Phaseolus vulgaris

          Stem-loop ID pvu-MIR159a
          Stem-loop Accession # MI0010700
          Stem-loop Sequence
          GUGGAGCUCCUUGAAGUCCAAUUGAGGAUCUUACUGGGUGGAUUGAGCUGCUUAGCUAUGGAUCCCACAGUUCUACCCAUCAGCAUGUUUUGUGGUAGUCUUGUGGCUUCCAUAUCUGGGGAGCUUCAUCUGCCUCUAUAGUAUCAUCCUUCUUUGGAUUGAAGGGAGCUCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015301
          miRBase ID: pvu-miR159a.1
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Populus trichocarpa

          Stem-loop ID ptc-MIR159a
          Stem-loop Accession # MI0002195
          Stem-loop Sequence
          GAUUAGGGAGUGGAGCUCCUUGAAGUCCAAUAGAGGUUCUUGCUGGGUAGAUUAAGCUGCUAAGCUAUGGAUCCACAGUCCUAUCUAUCAACCGAAGGAUAGGUUUGCGGCUUGCAUAUCUCAGGAGCUUUAUUGCCUAAUGUUAGAUCCCUUUUUGGAUUGAAGGGAGCUCUAAACCCAUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001901
          miRBase ID: ptc-miR159a
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Stem-loop ID ptc-MIR159b
          Stem-loop Accession # MI0002196
          Stem-loop Sequence
          GAUUAUGGAGUGGAGCUCCUUGAAGUCCAAUAGAAGCUCCUGCUGGGUAGAUCGAGCUGCUGAGCUAUGAAUCCCACAGCCCUAUCACCAUCAGUCAUUUUGAUGGGCCUGCGGCUUGCAUAUCUCAGGAGCUUUAUUACCUAAUGUUAGAUCUUUUUUUGGAUUGAAGGGAGCUCUAAACCUUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001902
          miRBase ID: ptc-miR159b
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Prunus persica

          Stem-loop ID ppe-MIR159
          Stem-loop Accession # MI0026065
          Stem-loop Sequence
          GGAGCAGAGCUCCUUGAAGUCCAAUAGAAGGUCUUGCUGGGUGGAUUGAGCUGCUGAGCUAUGGAUCCCACAGCCCUAUCUCAGAUCAUCAGAGGAAUAGGCUUGUGGCUUGCAUAUCUCAGGAGCUGCAUUACCUGUGUUAGAUCUUUGUUUGGAUUGAAGGGAGCUCUACACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031437
          miRBase ID: ppe-miR159
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Ricinus communis

          Stem-loop ID rco-MIR159
          Stem-loop Accession # MI0013378
          Stem-loop Sequence
          GAGUGGAGCUCCUUGAAGUCCAAUAGAGGAUCUUGAUGGGUAGAUUGAGCUGCUGAGCUAUGGAUCCCACAGCCCUAUAAGCAGAGAUUAUAAUUUUGGAUAGGCUUGUGGCUUGCAUAUCUCAGGAGCUUCAUUACCUUGUAUUAGAUCCUUGUUUGGAUUGAAGGGAGCUCUACAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014152
          miRBase ID: rco-miR159
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Solanum lycopersicum

          Stem-loop ID sly-MIR159
          Stem-loop Accession # MI0009974
          Stem-loop Sequence
          UGGAGCUCCUUGAAGUCCAACAAAAAAUCUAACAGGUUAAAUUGAGCUGCUGACCUAUGGAUUCCUCAGCCCUAUCUAUUUAUGAUUUCAAACAUAUAGAUAGGUUUUGGGUUUGCAUAUGUCAGGAGCUUUAUUUUACCCUUUGUUUGAUCAUUUUUUGGAUUGAAGGGAGCUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009141
          miRBase ID: sly-miR159
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Vitis vinifera

          Stem-loop ID vvi-MIR159c
          Stem-loop Accession # MI0006495
          Stem-loop Sequence
          UUUUGGAGUGGAGCUCCUUGAAGUCCAAUAGAGGGUCUUACUGGGUAGAUUGAGCUGCUGACUUAUGGAUCCCACAGCCCUAUCCCGUCAAUGGGGGGCAUUGGAUAGGCUUGUGGCUUGCAUAUCUCAGGAGCUGCAUUAUCCAAGUUUAGAUCCUUGUUUGGAUUGAAGGGAGCUCUACACCUCUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005650
          miRBase ID: vvi-miR159c
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA

          Vriesea carinata

          Stem-loop ID vca-MIR159
          Stem-loop Accession # MI0032981
          Stem-loop Sequence
          GUGGAGCUUCCUUGGGUCCAAAGAAGGGUCUGUUAGGGGCAAUUUUGCUGCUGGGUUAUGGAUCCCACUCUCCUAUCUCAUAGAAGAAAAGAAGAGAAAGAAAGAGUGGUAGAUGGAUUUGAGUAUUUGAGAUAGUUUUAGUGGCUUGCAUGACACAGGAGCUGCAUUAGCCCUUACAUGAUCUCUAUUUGGAUUGAAGGGAGCUCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040991
          miRBase ID: vca-miR159-3p
          Mature miRNA Sequence: UUUGGAUUGAAGGGAGCUCUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 000338
          mirVana® miRNA mimic : MC10332
          TaqMan™ Advanced miRNA Assay : 478411_mir


          miRBase Alias:

          Arabidopsis lyrata: aly-miR159a (v18)
          Citrus sinensis: csi-miR159 (v21)
          Glycine max: gma-miR159a (v17)
          Microbacterium esteraromaticum: mes-miR159 (v19)
          Microbacterium esteraromaticum: mes-miR159a (v21)
          Populus trichocarpa: ptc-miR159c (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline