Search
Search

| Assay Name: | hsa-miR-98-5p |
|---|---|
| miRBase Accession Number: | MI0000100 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGGUAGUAAGUUGUAUUGUU |
| Species: | Human, Mouse, Rat, Ateles geoffroyi, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Goat, Gorilla gorilla, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Ophiophagus hannah, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Tupaia chinensis, Xenopus laevis, Xenopus tropicalis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | UGAGGUAGUAAGUUGUAUUGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-98-5p | MIMAT0000096 |
| Mouse | mmu-miR-98-5p | MIMAT0000545 |
| Rat | rno-miR-98-5p | MIMAT0000819 |
| Ateles geoffroyi | age-miR-98 | MIMAT0002408 |
| Bovine | bta-miR-98 | MIMAT0003809 |
| Callithrix jacchus | cja-miR-98 | MIMAT0049302 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-98 | MIMAT0024015 |
| Dasypus novemcinctus | dno-miR-98-5p | MIMAT0047592 |
| Daubentonia madagascariensis | dma-miR-98 | MIMAT0049292 |
| Dog | cfa-miR-98 | MIMAT0006756 |
| Goat | chi-miR-98-5p | MIMAT0036310 |
| Gorilla gorilla | ggo-miR-98 | MIMAT0002406 |
| Guinea pig | cpo-miR-98-5p | MIMAT0046939 |
| Horse | eca-miR-98 | MIMAT0013247 |
| Microcebus murinus | mmr-miR-98 | MIMAT0049163 |
| Nomascus leucogenys | nle-miR-98 | MIMAT0049366 |
| Ophiophagus hannah | oha-miR-98-5p | MIMAT0036962 |
| Ornithorhynchus anatinus | oan-miR-98 | MIMAT0007178 |
| Otolemur garnettii | oga-miR-98 | MIMAT0049654 |
| Pan paniscus | ppa-miR-98 | MIMAT0002409 |
| Pan troglodytes | ptr-miR-98 | MIMAT0002405 |
| Papio hamadryas | pha-miR-98 | MIMAT0049569 |
| Pig | ssc-miR-98 | MIMAT0013905 |
| Pongo pygmaeus | ppy-miR-98 | MIMAT0002407 |
| Pteropus alecto | pal-miR-98-5p | MIMAT0039882 |
| Python bivittatus | pbv-miR-98-5p | MIMAT0038893 |
| Rabbit | ocu-miR-98-5p | MIMAT0048180 |
| Rhesus monkey | mml-miR-98 | MIMAT0002404 |
| Tupaia chinensis | tch-miR-98-5p | MIMAT0036570 |
| Xenopus laevis | xla-miR-98-5p | MIMAT0046617 |
| Xenopus tropicalis | xtr-miR-98 | MIMAT0003581 |