Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10456
          Assay Name: hsa-miR-192-5p
          miRBase Accession Number: MI0000234
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 64891137 - 64891246 [-] on Build GRCh38
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Species: Human, Mouse, Rat, Dog, Goat, Gorilla gorilla, Guinea pig, Horse, Nomascus leucogenys, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Tupaia chinensis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10456

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000063, mir-192

          Mature miRNA Sequence:

          CUGACCUAUGAAUUGACAGCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-192-5p MIMAT0000222
          Mouse mmu-miR-192-5p MIMAT0000517
          Rat rno-miR-192-5p MIMAT0000867
          Dog cfa-miR-192 MIMAT0006632
          Goat chi-miR-192-5p MIMAT0036024
          Gorilla gorilla ggo-miR-192 MIMAT0024119
          View More Rows
          Guinea pig cpo-miR-192-5p MIMAT0047068
          Horse eca-miR-192 MIMAT0013049
          Nomascus leucogenys nle-miR-192 MIMAT0049384
          Ornithorhynchus anatinus oan-miR-192-5p MIMAT0007016
          Otolemur garnettii oga-miR-192 MIMAT0049704
          Pan paniscus ppa-miR-192 MIMAT0049059
          Pan troglodytes ptr-miR-192 MIMAT0008057
          Pig ssc-miR-192 MIMAT0013910
          Pongo pygmaeus ppy-miR-192 MIMAT0015786
          Rhesus monkey mml-miR-192-5p MIMAT0006224
          Tupaia chinensis tch-miR-192-5p MIMAT0036616

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-192
          Stem-loop Accession # MI0000234
          Stem-loop Sequence
          GCCGAGACCGAGUGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUGCUCUCGUCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUGUUCGCCUCAAUGCCAGC
          Chromosome Location Chr. 11 - 64891137 - 64891246 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000222
          miRBase ID: hsa-miR-192-5p
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: Chr. 11 - 64891137 - 64891246 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-192
          Stem-loop Accession # MI0000551
          Stem-loop Sequence
          CGUGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUACUCUUUUCUCUCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUGUUCACC
          Chromosome Location Chr. 19 - 6264844 - 6264932 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000517
          miRBase ID: mmu-miR-192-5p
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: Chr. 19 - 6264844 - 6264932 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-192
          Stem-loop Accession # MI0000935
          Stem-loop Sequence
          GUCAAGAUGGAGUGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUACUCUGAUCUCGCCUCUGGCUGCCAGUUCCAUAGGUCACAGGUAUGUUCGCCUCAAUGCCAGC
          Chromosome Location Chr. 1 - 221634971 - 221635080 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000867
          miRBase ID: rno-miR-192-5p
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: Chr. 1 - 221634971 - 221635080 [+] on Build Rnor_6.0

          Dog

          Stem-loop ID cfa-mir-192
          Stem-loop Accession # MI0008031
          Stem-loop Sequence
          CUGACCUAUGAAUUGACAGCCAGUGCUCUCAUCUCUCCUCUGGCUGCCAAUUCCAUAGGUCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006632
          miRBase ID: cfa-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-192
          Stem-loop Accession # MI0030665
          Stem-loop Sequence
          CGAGUGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUGCUCUCGUGUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUGUUCGCCUCAAUGCCAGCCACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036024
          miRBase ID: chi-miR-192-5p
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-192
          Stem-loop Accession # MI0020666
          Stem-loop Sequence
          GAGACCGAGUGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUGCUCUCGUCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUGUUCGCCUCAAUGCCAGCCACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024119
          miRBase ID: ggo-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-192
          Stem-loop Accession # MI0038680
          Stem-loop Sequence
          CUGACCUAUGAAUUGACAGCCAGUGCUGUCAGCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047068
          miRBase ID: cpo-miR-192-5p
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-192
          Stem-loop Accession # MI0012792
          Stem-loop Sequence
          CUGACCUAUGAAUUGACAGCCAGUGCUCUUGUCUCCCUCUGGCUGCCAAUUCCAUAGGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013049
          miRBase ID: eca-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-192
          Stem-loop Accession # MI0040122
          Stem-loop Sequence
          CUGACCUAUGAAUUGACAGCCAGUGCUCUCAUCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049384
          miRBase ID: nle-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-192
          Stem-loop Accession # MI0006794
          Stem-loop Sequence
          AGAGCGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUUCUCUCUGACCCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUGUUCGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007016
          miRBase ID: oan-miR-192-5p
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-192
          Stem-loop Accession # MI0040488
          Stem-loop Sequence
          CUGACCUAUGAAUUGACAGCCAGUGCUCUCAUCUCCCCUCUGGCUGCCAAUUACAUAGGUCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049704
          miRBase ID: oga-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-192
          Stem-loop Accession # MI0039767
          Stem-loop Sequence
          CUGACCUAUGAAUUGACAGCCAGUGCUCUCGUCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049059
          miRBase ID: ppa-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-192
          Stem-loop Accession # MI0008565
          Stem-loop Sequence
          GCCGAGACCGAGUGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUGCUCUCGUCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUGUUCGCCUCAAUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008057
          miRBase ID: ptr-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-192
          Stem-loop Accession # MI0013127
          Stem-loop Sequence
          GCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUGCUCUCAUCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013910
          miRBase ID: ssc-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-192
          Stem-loop Accession # MI0014854
          Stem-loop Sequence
          GCCGAGACCGAGUGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUGCUCUCGUCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUGUUCGCCUCAAUGCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015786
          miRBase ID: ppy-miR-192
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-192
          Stem-loop Accession # MI0007654
          Stem-loop Sequence
          GCUGAGACCGAGUGCACAGGGCUCUGACCUAUGAAUUGACAGCCAGUGCUCUCGUCUCCCCUCUGGCUGCCAAUUCCAUAGGUCACAGGUAUGUUCGCCUCAAUGCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006224
          miRBase ID: mml-miR-192-5p
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-192
          Stem-loop Accession # MI0031287
          Stem-loop Sequence
          CUGACCUAUGAAUUGACAGCCAGUGCCCUCGUCUCUCCUCUGGCUGCCAAUUCCAUAGGUCACAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036616
          miRBase ID: tch-miR-192-5p
          Mature miRNA Sequence: CUGACCUAUGAAUUGACAGCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306714_pri
          TaqMan™ Pri-miRNA Assay : Hs03295148_pri
          TaqMan™ Pri-miRNA Assay : Rn03466059_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10456
          Ambion® Pre-miR™ miRNA Precursor : PM10456
          TaqMan™ MicroRNA Assay : 000491
          mirVana® miRNA mimic : MC10456
          TaqMan™ Advanced miRNA Assay : 478262_mir
          TaqMan™ Advanced miRNA Assay : rno480876_mir
          TaqMan™ Advanced miRNA Assay : mmu480876_mir


          miRBase Alias:

          Human: hsa-miR-192 (v17)
          Mouse: mmu-miR-192 (v17)
          Rat: rno-miR-192 (v18)
          Macaca mulatta: mml-miR-192 (v19)
          Ochrobactrum anthropi: oan-miR-192 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline