Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH11575
          Assay Name: hsa-miR-425-5p
          miRBase Accession Number: MI0001448
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 49020148 - 49020234 [-] on Build GRCh38
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Species: Human, Mouse, Rat, Cricetulus griseus, Dog, Goat, Monodelphis domestica, Ornithorhynchus anatinus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tupaia chinensis, Xenopus tropicalis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH11575

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000242, mir-425

          Mature miRNA Sequence:

          AAUGACACGAUCACUCCCGUUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-425-5p MIMAT0003393
          Mouse mmu-miR-425-5p MIMAT0004750
          Rat rno-miR-425-5p MIMAT0005314
          Cricetulus griseus cgr-miR-425-5p MIMAT0023958
          Dog cfa-miR-425 MIMAT0006639
          Goat chi-miR-425-5p MIMAT0036224
          View More Rows
          Monodelphis domestica mdo-miR-425-5p MIMAT0026697
          Ornithorhynchus anatinus oan-miR-425-5p MIMAT0007154
          Pan troglodytes ptr-miR-425 MIMAT0008144
          Pig ssc-miR-425-5p MIMAT0013917
          Pongo pygmaeus ppy-miR-425 MIMAT0015885
          Pteropus alecto pal-miR-425-5p MIMAT0039983
          Rhesus monkey mml-miR-425 MIMAT0006324
          Tupaia chinensis tch-miR-425-5p MIMAT0036572
          Xenopus tropicalis xtr-miR-425-5p MIMAT0003640

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-425
          Stem-loop Accession # MI0001448
          Stem-loop Sequence
          GAAAGCGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCACCCGAGAAGCCAUCGGGAAUGUCGUGUCCGCCCAGUGCUCUUUC
          Chromosome Location Chr. 3 - 49020148 - 49020234 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003393
          miRBase ID: hsa-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: Chr. 3 - 49020148 - 49020234 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-425
          Stem-loop Accession # MI0001447
          Stem-loop Sequence
          AAAGUGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCACCCAAGAAGCCAUCGGGAAUGUCGUGUCCGCCCAGUGCUCUUU
          Chromosome Location Chr. 9 - 108568777 - 108568861 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004750
          miRBase ID: mmu-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: Chr. 9 - 108568777 - 108568861 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-425
          Stem-loop Accession # MI0006146
          Stem-loop Sequence
          AGUGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCACCCAAGAAGCCAUCGGGAAUAUCGUGUCCGCCCAGUGCUCUUU
          Chromosome Location Chr. 8 - 117354821 - 117354903 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005314
          miRBase ID: rno-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: Chr. 8 - 117354821 - 117354903 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-425
          Stem-loop Accession # MI0020524
          Stem-loop Sequence
          GUGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCACCCAAGAAGCCAUCGGGAAUGUCGUGUCCGCCCAGUGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023958
          miRBase ID: cgr-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-425
          Stem-loop Accession # MI0008038
          Stem-loop Sequence
          AAUGACACGAUCACUCCCGUUGAGUGGGCACCCAAGAAGCCAUCGGGAAUGUCGUGUCCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006639
          miRBase ID: cfa-miR-425
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-425
          Stem-loop Accession # MI0030790
          Stem-loop Sequence
          AAAGUGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCACCCAAGAAGCCAUCGGGAAUGUCGUGUCCGCCCAGUGCUCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036224
          miRBase ID: chi-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-425
          Stem-loop Accession # MI0005342
          Stem-loop Sequence
          AAAGUGCUUUGGAAUGACACGAUCACUCCCGUUGAGCGGACAGCCAAGAAGCCAUCGGGAAUAUCGUGUCCGUCCAAUGCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026697
          miRBase ID: mdo-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-425
          Stem-loop Accession # MI0006880
          Stem-loop Sequence
          GGGAGAGAAAUCGCUUUGGAAUGACACGAUCACUCCCGUUGAGCGGACAGCCAAGAAGCCAUCGGGAAUAUCGUGUCCGUCCAAAGCUCUUUCGGGGCCUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007154
          miRBase ID: oan-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-425
          Stem-loop Accession # MI0008665
          Stem-loop Sequence
          GAAAGCGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCACCCGAGAAGCCAUCGGGAAUGUCGUGUCCGCCCAGUGCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008144
          miRBase ID: ptr-miR-425
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-425
          Stem-loop Accession # MI0013133
          Stem-loop Sequence
          AAGUGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCACCCAAGUAGCCAUCGGGAAUGUCGUGUCCGCCCAGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013917
          miRBase ID: ssc-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-425
          Stem-loop Accession # MI0014941
          Stem-loop Sequence
          GAAAGCGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCACCCGAGAAGCCAUCGGGAAUGUCGUGUCCGCCCAGUGCUCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015885
          miRBase ID: ppy-miR-425
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-425
          Stem-loop Accession # MI0032433
          Stem-loop Sequence
          AAUGACACGAUCACUCCCGUUGAGUGGGCACCCAAGAAGCCAUCGGGAAUGUCGUGUCCGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039983
          miRBase ID: pal-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-425
          Stem-loop Accession # MI0007741
          Stem-loop Sequence
          GAAAGCGCUUUGGAAUGACACGAUCACUCCCGUUGAGUGGGCCCCCGAGAAGCCAUCGGGAAUGUCGUGUCCGCCCAGUGCUCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006324
          miRBase ID: mml-miR-425
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-425
          Stem-loop Accession # MI0031235
          Stem-loop Sequence
          AAUGACACGAUCACUCCCGUUGAGUGGGCACCCAAGAAGCCAUCGGGAAUGUCGUGUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036572
          miRBase ID: tch-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-425
          Stem-loop Accession # MI0004882
          Stem-loop Sequence
          GCCUUGGAAUGACACGAUCACUCCCGUUGAGCCGAAACUCACAGCGCCAUCGGGAAUAUCGUGUCGCUCCAAAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003640
          miRBase ID: xtr-miR-425-5p
          Mature miRNA Sequence: AAUGACACGAUCACUCCCGUUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303194_pri
          TaqMan™ Pri-miRNA Assay : Rn03465415_pri
          TaqMan™ Pri-miRNA Assay : Mm03307466_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11575
          Ambion® Pre-miR™ miRNA Precursor : PM11575
          TaqMan™ MicroRNA Assay : 001516
          mirVana® miRNA mimic : MC11575
          TaqMan™ Advanced miRNA Assay : 478094_mir
          TaqMan™ Advanced miRNA Assay : rno481161_mir
          TaqMan™ Advanced miRNA Assay : mmu481161_mir


          miRBase Alias:

          Human: hsa-miR-425 (v17)
          Mouse: mmu-miR-425 (v17)
          Rat: rno-miR-425 (v18)
          Ochrobactrum anthropi: oan-miR-425 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline