Search
Search

| Assay Name: | pma-miR-1a-3p |
|---|---|
| miRBase Accession Number: | MI0016999 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAAUGUAAAGAAGUACGUUG |
| Species: | Petromyzon marinus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000038, mir-1 |
Mature miRNA Sequence: | UGGAAUGUAAAGAAGUACGUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Petromyzon marinus | pma-miR-1a-3p | MIMAT0019365 |