Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11749
          Assay Name: hsa-miR-139-5p
          miRBase Accession Number: MI0000261
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 72615063 - 72615130 [-] on Build GRCh38
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Species: Human, Bovine, Callithrix jacchus, Dasypus novemcinctus, Rabbit, Tupaia chinensis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11749
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000117, mir-139

          Mature miRNA Sequence:

          UCUACAGUGCACGUGUCUCCAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-139-5p MIMAT0000250
          Bovine bta-miR-139 MIMAT0003788
          Callithrix jacchus cja-miR-139 MIMAT0039472
          Dasypus novemcinctus dno-miR-139-5p MIMAT0047651
          Rabbit ocu-miR-139-5p MIMAT0048239
          Tupaia chinensis tch-miR-139-5p MIMAT0036540

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-139
          Stem-loop Accession # MI0000261
          Stem-loop Sequence
          GUGUAUUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAAC
          Chromosome Location Chr. 11 - 72615063 - 72615130 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000250
          miRBase ID: hsa-miR-139-5p
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: Chr. 11 - 72615063 - 72615130 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-139
          Stem-loop Accession # MI0005009
          Stem-loop Sequence
          GUGUACUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003788
          miRBase ID: bta-miR-139
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-139
          Stem-loop Accession # MI0031996
          Stem-loop Sequence
          CUGGCUCAGGUGUAUUCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGUAACAACUGAAGCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039472
          miRBase ID: cja-miR-139
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-139
          Stem-loop Accession # MI0038988
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUCGGAGGCUGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047651
          miRBase ID: dno-miR-139-5p
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-139
          Stem-loop Accession # MI0039297
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUUGGAGGCUGGAGACGCGGCCCUGUUGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048239
          miRBase ID: ocu-miR-139-5p
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-139
          Stem-loop Accession # MI0031199
          Stem-loop Sequence
          UCUACAGUGCACGUGUCUCCAGUGUGGCUCAGAGGCUGGAGACGCGGCCCUGUCGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036540
          miRBase ID: tch-miR-139-5p
          Mature miRNA Sequence: UCUACAGUGCACGUGUCUCCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302845_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11749
          TaqMan™ MicroRNA Assay : 005364_mat
          mirVana® miRNA inhibitor : MH11749
          mirVana® miRNA mimic : MC11749
          TaqMan™ Advanced miRNA Assay : 478312_mir

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline