Search
Search

| Assay Name: | hsa-miR-29a |
|---|---|
| miRBase Accession Number: | MI0020478 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGCACCAUCUGAAAUCGGUU |
| Species: | Cricetulus griseus, Goat, Sheep, Tupaia chinensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | UAGCACCAUCUGAAAUCGGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-29a-3p | MIMAT0023884 |
| Goat | chi-miR-29a-3p | MIMAT0036113 |
| Sheep | oar-miR-29a | MIMAT0014967 |
| Tupaia chinensis | tch-miR-29a-3p | MIMAT0036511 |