Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10932
          Assay Name: cgr-miR-29a-3p
          miRBase Accession Number: MI0020478
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUU
          Species: Cricetulus griseus, Goat, Sheep, Tupaia chinensis
          Product Type: mirVana® miRNA mimic
          Assay ID MC10932

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000009, mir-29

          Mature miRNA Sequence:

          UAGCACCAUCUGAAAUCGGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Cricetulus griseus cgr-miR-29a-3p MIMAT0023884
          Goat chi-miR-29a-3p MIMAT0036113
          Sheep oar-miR-29a MIMAT0014967
          Tupaia chinensis tch-miR-29a-3p MIMAT0036511

          Stem-loop Details



          Cricetulus griseus

          Stem-loop ID cgr-mir-29a
          Stem-loop Accession # MI0020478
          Stem-loop Sequence
          UUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAGAAUUUUCUAGCACCAUCUGAAAUCGGUUAUAAUGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023884
          miRBase ID: cgr-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-29a
          Stem-loop Accession # MI0030726
          Stem-loop Sequence
          CCCCUUAGAGGAUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAUAAUGAUUGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036113
          miRBase ID: chi-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-29a
          Stem-loop Accession # MI0014117
          Stem-loop Sequence
          AUGACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014967
          miRBase ID: oar-miR-29a
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-29a
          Stem-loop Accession # MI0031169
          Stem-loop Sequence
          ACUGAUUUCUUUUGGUGUUCAGAGUCAAUAUAAUUUUCUAGCACCAUCUGAAAUCGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036511
          miRBase ID: tch-miR-29a-3p
          Mature miRNA Sequence: UAGCACCAUCUGAAAUCGGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10932
          Ambion® Pre-miR™ miRNA Precursor : PM10932
          TaqMan™ MicroRNA Assay : 000412
          mirVana® miRNA inhibitor : MH10932

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline