Search
Search

| Assay Name: | mmu-miR-322* |
|---|---|
| miRBase Accession Number: | MI0000590 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAACAUGAAGCGCUGCAACAC |
| Species: | Mouse, Cricetulus griseus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000164, mir-322 |
Mature miRNA Sequence: | AAACAUGAAGCGCUGCAACAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-322-3p | MIMAT0000549 |
| Cricetulus griseus | cgr-miR-322-3p | MIMAT0023908 |