Search
Search

| Assay Name: | ame-miR-31a |
|---|---|
| miRBase Accession Number: | MI0005737 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGCAAGAUGUCGGCAUAGCUGA |
| Species: | Apis mellifera, Nasonia giraulti, Nasonia longicornis, Nasonia vitripennis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | GGCAAGAUGUCGGCAUAGCUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Apis mellifera | ame-miR-31a-5p | MIMAT0004428 |
| Nasonia giraulti | ngi-miR-31a | MIMAT0018387 |
| Nasonia longicornis | nlo-miR-31a | MIMAT0018434 |
| Nasonia vitripennis | nvi-miR-31a | MIMAT0015661 |