Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12210
          Assay Name: ame-miR-31a-5p
          miRBase Accession Number: MI0005737
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: GGCAAGAUGUCGGCAUAGCUGA
          Species: Apis mellifera, Nasonia giraulti, Nasonia longicornis, Nasonia vitripennis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12210

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000064, mir-31

          Mature miRNA Sequence:

          GGCAAGAUGUCGGCAUAGCUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Apis mellifera ame-miR-31a-5p MIMAT0004428
          Nasonia giraulti ngi-miR-31a MIMAT0018387
          Nasonia longicornis nlo-miR-31a MIMAT0018434
          Nasonia vitripennis nvi-miR-31a MIMAT0015661

          Stem-loop Details



          Apis mellifera

          Stem-loop ID ame-mir-31a
          Stem-loop Accession # MI0005737
          Stem-loop Sequence
          AUCACGAUUCUAACUGGGCGCCUCGAAGGCAAGAUGUCGGCAUAGCUGAUGCGAUUUUAAAAUUCGGCUGUGUCACAUCCAGCCAACCGAACGCUCAGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004428
          miRBase ID: ame-miR-31a-5p
          Mature miRNA Sequence: GGCAAGAUGUCGGCAUAGCUGA
          Chromosome Location: NA

          Nasonia giraulti

          Stem-loop ID ngi-mir-31a
          Stem-loop Accession # MI0016628
          Stem-loop Sequence
          GAAGGCAAGAUGUCGGCAUAGCUGAUGCUUUUGAAAUUUCGGCUGUGUCACAUCCAGCCAGCCAAACGCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018387
          miRBase ID: ngi-miR-31a
          Mature miRNA Sequence: GGCAAGAUGUCGGCAUAGCUGA
          Chromosome Location: NA

          Nasonia longicornis

          Stem-loop ID nlo-mir-31a
          Stem-loop Accession # MI0016675
          Stem-loop Sequence
          GAAGGCAAGAUGUCGGCAUAGCUGAUGCUUUUGAAAUUUCGGCUGUGUCACAUCCAGCCAGCCAAACGCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018434
          miRBase ID: nlo-miR-31a
          Mature miRNA Sequence: GGCAAGAUGUCGGCAUAGCUGA
          Chromosome Location: NA

          Nasonia vitripennis

          Stem-loop ID nvi-mir-31a
          Stem-loop Accession # MI0014722
          Stem-loop Sequence
          UGCAGCGUAAAAUAUGAGCGUGUAGAAGGCAAGAUGUCGGCAUAGCUGAUGCUUUUGAAAUUUCGGCUGUGUCACAUCCAGCCAGCCAAACGCUCAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015661
          miRBase ID: nvi-miR-31a
          Mature miRNA Sequence: GGCAAGAUGUCGGCAUAGCUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 007632_mat
          mirVana® miRNA mimic : MC12210


          miRBase Alias:

          Anaerococcus mediterraneensis: ame-miR-31a (v21)
          Trichosporon caseorum: tca-miR-31a (v16)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline