Search
Search

| Assay Name: | hsa-miR-32-3p |
|---|---|
| Stem-loop Accession Number: | MI0000090 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAUUUAGUGUGUGUGAUAUUU |
| Species: | Human, Ophiophagus hannah |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000069, mir-32 |
Mature miRNA Sequence: | CAAUUUAGUGUGUGUGAUAUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-32-3p | MIMAT0004505 |
| Ophiophagus hannah | oha-miR-32-3p | MIMAT0036900 |