Search
Search

| Assay Name: | hsa-miR-660-5p |
|---|---|
| miRBase Accession Number: | MI0003684 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGUUG |
| Species: | Human, Callithrix jacchus, Daubentonia madagascariensis, Dog, Gorilla gorilla, Horse, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | UACCCAUUGCAUAUCGGAGUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-660-5p | MIMAT0003338 |
| Callithrix jacchus | cja-miR-660 | MIMAT0039432 |
| Daubentonia madagascariensis | dma-miR-660 | MIMAT0049268 |
| Dog | cfa-miR-660 | MIMAT0006760 |
| Gorilla gorilla | ggo-miR-660 | MIMAT0024164 |
| Horse | eca-miR-660 | MIMAT0013241 |
| View More Rows | ||
| Pan paniscus | ppa-miR-660 | MIMAT0049103 |
| Pan troglodytes | ptr-miR-660 | MIMAT0008308 |
| Papio hamadryas | pha-miR-660 | MIMAT0049544 |
| Pig | ssc-miR-660 | MIMAT0041612 |
| Pongo pygmaeus | ppy-miR-660 | MIMAT0016092 |
| Pteropus alecto | pal-miR-660-5p | MIMAT0039997 |
| Rhesus monkey | mml-miR-660-5p | MIMAT0006499 |