Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs03304909_pri
Assay Name:
hsa-mir-660
Stem-loop ID:
hsa-mir-660
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.X: 50013241 - 50013337 [+] on Build GRCh38
Stem-loop Location:
Chr.X: 50013241 - 50013337 [+] on Build GRCh38
Mature miRNA ID:
hsa-miR-660-5p
hsa-miR-660-3p
Species:
Human
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Hs03304909_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
66
Chromosome Location:
Chr. X - 50013542 - 50013542 [-] on 50013542 Build GRCh38
Species:
Human
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Human
hsa-miR-660-5p
MIMAT0003338
Human
hsa-miR-660-3p
MIMAT0022711
Stem-loop Details
Human
Stem-loop ID
hsa-mir-660
Stem-loop Accession #
MI0003684
Stem-loop Sequence
CUGCUCCUUCUCCCAUACCCAUUGCAUAUCGGAGUUGUGAAUUCUCAAAACACCUCCUGUGUGCAUGGAUUACAGGAGGGUGAGCCUUGUCAUCGUG
Chromosome Location
Chr. X - 50013241 - 50013337 [+] on Build GRCh38
Mature MicroRNA
miRBase Accession #: MIMAT0003338
miRBase ID: hsa-miR-660-5p
Mature miRNA Sequence: UACCCAUUGCAUAUCGGAGUUG
Chromosome Location: Chr. X - 50013241 - 50013337 [+] on Build GRCh38
miRBase Accession #: MIMAT0022711
miRBase ID: hsa-miR-660-3p
Mature miRNA Sequence: ACCUCCUGUGUGCAUGGAUUA
Chromosome Location: Chr. X - 50013241 - 50013337 [+] on Build GRCh38
Gene Family ID
MIPF0000113,
mir-188
Distance to Amplicon
173 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM11216
Ambion
®
Pre-miR
™
miRNA Precursor :
PM11216
TaqMan
™
MicroRNA Assay :
001515
mir
Vana
®
miRNA inhibitor :
MH11216
mir
Vana
®
miRNA mimic :
MC11216
TaqMan
™
Advanced miRNA Assay :
478192_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM23930
Ambion
®
Pre-miR
™
miRNA Precursor :
PM23930
TaqMan
™
MicroRNA Assay :
470860_mat
mir
Vana
®
miRNA inhibitor :
MH23930
mir
Vana
®
miRNA mimic :
MC23930
TaqMan
™
Advanced miRNA Assay :
479143_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline