Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs03303803_pri
Assay Name:
hsa-mir-484
Stem-loop ID:
hsa-mir-484
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.16: 15643294 - 15643372 [+] on Build GRCh38
Stem-loop Location:
Chr.16: 15643294 - 15643372 [+] on Build GRCh38
Mature miRNA ID:
hsa-miR-484
Species:
Human
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Hs03303803_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
58
Chromosome Location:
Chr. 16 - 15642992 - 15642992 [-] on 15642992 Build GRCh38
Species:
Human
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Human
hsa-miR-484
MIMAT0002174
Stem-loop Details
Human
Stem-loop ID
hsa-mir-484
Stem-loop Accession #
MI0002468
Stem-loop Sequence
AGCCUCGUCAGGCUCAGUCCCCUCCCGAUAAACCCCUAAAUAGGGACUUUCCCGGGGGGUGACCCUGGCUUUUUUGGCG
Chromosome Location
Chr. 16 - 15643294 - 15643372 [+] on Build GRCh38
Mature MicroRNA
miRBase Accession #: MIMAT0002174
miRBase ID: hsa-miR-484
Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
Chromosome Location: Chr. 16 - 15643294 - 15643372 [+] on Build GRCh38
Gene Family ID
MIPF0000219,
mir-484
Distance to Amplicon
272 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10379
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10379
TaqMan
™
MicroRNA Assay :
001821
mir
Vana
®
miRNA inhibitor :
MH10379
mir
Vana
®
miRNA mimic :
MC10379
TaqMan
™
Advanced miRNA Assay :
rno481181_mir
TaqMan
™
Advanced miRNA Assay :
478308_mir
TaqMan
™
Advanced miRNA Assay :
mmu481181_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline