Search
Search

| Assay Name: | hsa-miR-484 |
|---|---|
| miRBase Accession Number: | MI0002468 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAGGCUCAGUCCCCUCCCGAU |
| Species: | Human, Mouse, Rat, Bovine, Cricetulus griseus, Nomascus leucogenys, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000219, mir-484 |
Mature miRNA Sequence: | UCAGGCUCAGUCCCCUCCCGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-484 | MIMAT0002174 |
| Mouse | mmu-miR-484 | MIMAT0003127 |
| Rat | rno-miR-484 | MIMAT0005319 |
| Bovine | bta-miR-484 | MIMAT0003535 |
| Cricetulus griseus | cgr-miR-484 | MIMAT0023966 |
| Nomascus leucogenys | nle-miR-484 | MIMAT0049338 |
| View More Rows | ||
| Pan paniscus | ppa-miR-484 | MIMAT0049105 |
| Pan troglodytes | ptr-miR-484 | MIMAT0008158 |
| Pongo pygmaeus | ppy-miR-484 | MIMAT0015901 |
| Rhesus monkey | mml-miR-484 | MIMAT0006341 |