Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10379
          Assay Name: hsa-miR-484
          miRBase Accession Number: MI0002468
          miRBase Version: v22.1
          Chromosome Location: Chr.16: 15643294 - 15643372 [+] on Build GRCh38
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Species: Human, Mouse, Rat, Bovine, Cricetulus griseus, Nomascus leucogenys, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10379
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000219, mir-484

          Mature miRNA Sequence:

          UCAGGCUCAGUCCCCUCCCGAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-484 MIMAT0002174
          Mouse mmu-miR-484 MIMAT0003127
          Rat rno-miR-484 MIMAT0005319
          Bovine bta-miR-484 MIMAT0003535
          Cricetulus griseus cgr-miR-484 MIMAT0023966
          Nomascus leucogenys nle-miR-484 MIMAT0049338
          View More Rows
          Pan paniscus ppa-miR-484 MIMAT0049105
          Pan troglodytes ptr-miR-484 MIMAT0008158
          Pongo pygmaeus ppy-miR-484 MIMAT0015901
          Rhesus monkey mml-miR-484 MIMAT0006341

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-484
          Stem-loop Accession # MI0002468
          Stem-loop Sequence
          AGCCUCGUCAGGCUCAGUCCCCUCCCGAUAAACCCCUAAAUAGGGACUUUCCCGGGGGGUGACCCUGGCUUUUUUGGCG
          Chromosome Location Chr. 16 - 15643294 - 15643372 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002174
          miRBase ID: hsa-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: Chr. 16 - 15643294 - 15643372 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-484
          Stem-loop Accession # MI0003491
          Stem-loop Sequence
          CUCGUCAGGCUCAGUCCCCUCCCGAUAAACCUCAAAAUAGGGUCUUACCUAGGGGGCUGGCGGCGUA
          Chromosome Location Chr. 16 - 14159626 - 14159692 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003127
          miRBase ID: mmu-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: Chr. 16 - 14159626 - 14159692 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-484
          Stem-loop Accession # MI0006151
          Stem-loop Sequence
          GCAUGCAGGGAAGGGGGGGCGGGGCCUCGCGGCCCUGCUGCCUCGUCAGGCUCAGUCCCCUCCCGAUAAACCUCAAA
          Chromosome Location Chr. 10 - 27845 - 27921 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005319
          miRBase ID: rno-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: Chr. 10 - 27845 - 27921 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-484
          Stem-loop Accession # MI0004749
          Stem-loop Sequence
          GUCAGGCUCAGUCCCCUCCCGAUAAACCUCUAAAUAGGGACCUUCCCGGGGGGCUACCUCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003535
          miRBase ID: bta-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-484
          Stem-loop Accession # MI0020529
          Stem-loop Sequence
          UGCAGCCUCGUCAGGCUCAGUCCCCUCCCGAUAAACCUCUAAAUAGGGUGUUUCCCCGGGGGUGGCCUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023966
          miRBase ID: cgr-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-484
          Stem-loop Accession # MI0040074
          Stem-loop Sequence
          UCAGGCUCAGUCCCCUCCCGAUAAACCCCUAAAUAGGGACUUUCCCGGGGGGUGACCCUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049338
          miRBase ID: nle-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-484
          Stem-loop Accession # MI0039817
          Stem-loop Sequence
          UCAGGCUCAGUCCCCUCCCGAUAAACCCCUAAAUAGGGACUUUCCCGGGGGGUGACCCUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049105
          miRBase ID: ppa-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-484
          Stem-loop Accession # MI0008680
          Stem-loop Sequence
          GCCUCGUCAGGCUCAGUCCCCUCCCGAUAAACCCCUAAAUAGGGACUUUCCCGGGGGGUGACCCUGGCUUUUUUGGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008158
          miRBase ID: ptr-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-484
          Stem-loop Accession # MI0014956
          Stem-loop Sequence
          AGCCUCGUCAGGCUCAGUCCCCUCCCGAUAAACCCCUAAAUAGGGACUUUCCCGGGGGGUGACCCUGGCUUUUUUGGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015901
          miRBase ID: ppy-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-484
          Stem-loop Accession # MI0007757
          Stem-loop Sequence
          GUCAGGCUCAGUCCCCUCCCGAUAAACCCCUAAAUAGGGACUUUCCCGGGGGGUGACCCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006341
          miRBase ID: mml-miR-484
          Mature miRNA Sequence: UCAGGCUCAGUCCCCUCCCGAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303803_pri
          TaqMan™ Pri-miRNA Assay : Rn03465173_pri
          TaqMan™ Pri-miRNA Assay : Mm03307626_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10379
          TaqMan™ MicroRNA Assay : 001821
          mirVana® miRNA inhibitor : MH10379
          mirVana® miRNA mimic : MC10379
          TaqMan™ Advanced miRNA Assay : rno481181_mir
          TaqMan™ Advanced miRNA Assay : 478308_mir
          TaqMan™ Advanced miRNA Assay : mmu481181_mir

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline