Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10245
          Assay Name: age-miR-124a
          miRBase Accession Number: MI0002762
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Species: Ateles geoffroyi, Chicken, Gorilla gorilla, Lagothrix lagotricha, Monodelphis domestica, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Xenopus tropicalis
          Product Type: mirVana® miRNA mimic
          Assay ID MC10245

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000021, mir-124

          Mature miRNA Sequence:

          UUAAGGCACGCGGUGAAUGCCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Ateles geoffroyi age-miR-124a MIMAT0002466
          Chicken gga-miR-124a-3p MIMAT0001128
          Gorilla gorilla ggo-miR-124a MIMAT0002465
          Lagothrix lagotricha lla-miR-124a MIMAT0002471
          Monodelphis domestica mdo-miR-124a-3p MIMAT0004102
          Pan paniscus ppa-miR-124a MIMAT0002467
          View More Rows
          Pan troglodytes ptr-miR-124a MIMAT0002469
          Pongo pygmaeus ppy-miR-124a MIMAT0002468
          Rhesus monkey mml-miR-124a-3p MIMAT0002470
          Xenopus tropicalis xtr-miR-124 MIMAT0003683

          Stem-loop Details



          Ateles geoffroyi

          Stem-loop ID age-mir-124a
          Stem-loop Accession # MI0002762
          Stem-loop Sequence
          AUCAAGAUCAGAGGCUCUGCCCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002466
          miRBase ID: age-miR-124a
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-124a
          Stem-loop Accession # MI0001197
          Stem-loop Sequence
          AGGCUCUGCCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAUCCUCCAGGCGGCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001128
          miRBase ID: gga-miR-124a-3p
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Stem-loop ID gga-mir-124a-2
          Stem-loop Accession # MI0008210
          Stem-loop Sequence
          CCGAGCCUCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGAGCCUCACAGCUUCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001128
          miRBase ID: gga-miR-124a-3p
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-124a
          Stem-loop Accession # MI0002761
          Stem-loop Sequence
          AUCAAGAUUAGAGGCUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCGCUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002465
          miRBase ID: ggo-miR-124a
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-124a
          Stem-loop Accession # MI0002767
          Stem-loop Sequence
          AUCAAGAUCAGAGGCUCUGCCCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGACUGCACUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002471
          miRBase ID: lla-miR-124a
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-124a-1
          Stem-loop Accession # MI0005288
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004102
          miRBase ID: mdo-miR-124a-3p
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Stem-loop ID mdo-mir-124a-2
          Stem-loop Accession # MI0005289
          Stem-loop Sequence
          AUCAGAGACUCUGUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004102
          miRBase ID: mdo-miR-124a-3p
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Stem-loop ID mdo-mir-124a-3
          Stem-loop Accession # MI0005290
          Stem-loop Sequence
          CUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004102
          miRBase ID: mdo-miR-124a-3p
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-124a
          Stem-loop Accession # MI0002763
          Stem-loop Sequence
          AUCAAGAUUAGAGGCUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002467
          miRBase ID: ppa-miR-124a
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-124a
          Stem-loop Accession # MI0002765
          Stem-loop Sequence
          AUCAAGAUUAGAGGCUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002469
          miRBase ID: ptr-miR-124a
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-124a
          Stem-loop Accession # MI0002764
          Stem-loop Sequence
          AUCAAGAUUAGAGGCUCUGCCCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002468
          miRBase ID: ppy-miR-124a
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-124a-1
          Stem-loop Accession # MI0002766
          Stem-loop Sequence
          AUCAAGAUCAGAGGCUCUGCCCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002470
          miRBase ID: mml-miR-124a-3p
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Stem-loop ID mml-mir-124a-2
          Stem-loop Accession # MI0007614
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002470
          miRBase ID: mml-miR-124a-3p
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-124
          Stem-loop Accession # MI0004930
          Stem-loop Sequence
          UAAGUCUCUGACUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGUGGAGCCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003683
          miRBase ID: xtr-miR-124
          Mature miRNA Sequence: UUAAGGCACGCGGUGAAUGCCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10245
          Ambion® Pre-miR™ miRNA Precursor : PM10245
          TaqMan™ MicroRNA Assay : 000446
          mirVana® miRNA inhibitor : MH10245


          miRBase Alias:

          Gallus gallus: gga-miR-124a (v19)
          Macaca mulatta: mml-miR-124a (v19)
          Monodelphis domestica: mdo-miR-124a (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline