Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10711
          Assay Name: mmu-miR-23b-3p
          miRBase Accession Number: MI0000141
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 63300484 - 63300557 [+] on Build GRCm38
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Species: Mouse, Rat, Anolis carolinensis, Astatotilapia burtoni, Callithrix jacchus, Chicken, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Daubentonia madagascariensis, Goat, Horse, Ictalurus punctatus, Metriaclima zebra, Microcebus murinus, Monodelphis domestica, Neolamprologus brichardi, Nomascus leucogenys, Oreochromis niloticus, Papio hamadryas, Pundamilia nyererei, Rhesus monkey, Taeniopygia guttata, Xenopus laevis, Xenopus tropicalis
          Product Type: mirVana® miRNA mimic
          Assay ID MC10711

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000027, mir-23

          Mature miRNA Sequence:

          AUCACAUUGCCAGGGAUUACC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-23b-3p MIMAT0000125
          Rat rno-miR-23b-3p MIMAT0000793
          Anolis carolinensis aca-miR-23b-3p MIMAT0021898
          Astatotilapia burtoni abu-miR-23b MIMAT0042134
          Callithrix jacchus cja-miR-23b MIMAT0039380
          Chicken gga-miR-23b-3p MIMAT0001186
          View More Rows
          Chrysemys picta cpi-miR-23b-3p MIMAT0037656
          Cricetulus griseus cgr-miR-23b-3p MIMAT0023865
          Cyprinus carpio ccr-miR-23b MIMAT0026278
          Daubentonia madagascariensis dma-miR-23b MIMAT0049279
          Goat chi-miR-23b-3p MIMAT0036089
          Horse eca-miR-23b MIMAT0013113
          Ictalurus punctatus ipu-miR-23b MIMAT0029482
          Metriaclima zebra mze-miR-23b MIMAT0042354
          Microcebus murinus mmr-miR-23b MIMAT0049228
          Monodelphis domestica mdo-miR-23b-3p MIMAT0004176
          Neolamprologus brichardi nbr-miR-23b MIMAT0042535
          Nomascus leucogenys nle-miR-23b MIMAT0049446
          Oreochromis niloticus oni-miR-23b MIMAT0042713
          Papio hamadryas pha-miR-23b MIMAT0049526
          Pundamilia nyererei pny-miR-23b MIMAT0042924
          Rhesus monkey mml-miR-23b-3p MIMAT0006165
          Taeniopygia guttata tgu-miR-23-3p MIMAT0014566
          Xenopus laevis xla-miR-23b-3p MIMAT0046549
          Xenopus tropicalis xtr-miR-23b MIMAT0003673

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-23b
          Stem-loop Accession # MI0000141
          Stem-loop Sequence
          GGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUGAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACC
          Chromosome Location Chr. 13 - 63300484 - 63300557 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000125
          miRBase ID: mmu-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: Chr. 13 - 63300484 - 63300557 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-23b
          Stem-loop Accession # MI0000853
          Stem-loop Sequence
          CUCACCUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUGAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCAUGACCUUGGC
          Chromosome Location Chr. 17 - 823222 - 823318 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000793
          miRBase ID: rno-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: Chr. 17 - 823222 - 823318 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-23b
          Stem-loop Accession # MI0018832
          Stem-loop Sequence
          GUUUUGGUUGUUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUGAAAAUCACAUUGCCAGGGAUUACCACAUAACCAUGACAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021898
          miRBase ID: aca-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-23b
          Stem-loop Accession # MI0033847
          Stem-loop Sequence
          AGGGUUCCUGGCGUGCUGAUUUGUGACUUAUGAUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042134
          miRBase ID: abu-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-23b
          Stem-loop Accession # MI0031898
          Stem-loop Sequence
          UGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039380
          miRBase ID: cja-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-23b
          Stem-loop Accession # MI0001273
          Stem-loop Sequence
          UGUUGUGGCUGUUUGGGUUCCUGGCAUGAUGAUUUGUGAGUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACAUAGCCAUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001186
          miRBase ID: gga-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-23b
          Stem-loop Accession # MI0029412
          Stem-loop Sequence
          UAAUGUUGUGGCUGUUUGGGUUCCUGGCAUGCUGAUUUGUGAUUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACAUAACCAUGACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037656
          miRBase ID: cpi-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-23b
          Stem-loop Accession # MI0020467
          Stem-loop Sequence
          CUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUGAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023865
          miRBase ID: cgr-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-23b
          Stem-loop Accession # MI0023379
          Stem-loop Sequence
          UGUGUGGGUUCCUGGCAUGCUGAUUUGUGACUGUAGUGAAAAAUCACAUUGCCAGGGAUUACCACACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026278
          miRBase ID: ccr-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-23b
          Stem-loop Accession # MI0040010
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049279
          miRBase ID: dma-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-23b
          Stem-loop Accession # MI0030712
          Stem-loop Sequence
          GUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCACGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036089
          miRBase ID: chi-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-23b
          Stem-loop Accession # MI0012863
          Stem-loop Sequence
          GGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013113
          miRBase ID: eca-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-23b-1
          Stem-loop Accession # MI0024595
          Stem-loop Sequence
          GGGUUCUUGGCAUGCUGAUUUGUGACUGUAGUAAAAAAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029482
          miRBase ID: ipu-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Stem-loop ID ipu-mir-23b-2
          Stem-loop Accession # MI0024594
          Stem-loop Sequence
          GGGUUCCUGGCGUGCUGAUUUGUGACUUAAGAUAAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029482
          miRBase ID: ipu-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-23b
          Stem-loop Accession # MI0034126
          Stem-loop Sequence
          AGGGUUCCUGGCGUGCUGAUUUGUGACUUAUGAUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042354
          miRBase ID: mze-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-23b
          Stem-loop Accession # MI0039955
          Stem-loop Sequence
          UUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049228
          miRBase ID: mmr-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-23b
          Stem-loop Accession # MI0005366
          Stem-loop Sequence
          GGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUGAAAAUCACAUUGCCAGGGAUUACCACGCAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004176
          miRBase ID: mdo-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-23b
          Stem-loop Accession # MI0034381
          Stem-loop Sequence
          AGGGUUCCUGGCGUGCUGAUUUGUGACUUAUGAUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042535
          miRBase ID: nbr-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-23b
          Stem-loop Accession # MI0040194
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049446
          miRBase ID: nle-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-23b
          Stem-loop Accession # MI0034627
          Stem-loop Sequence
          GGGUUCCUGGCGUGCUGAUUUGUGACUUAUGAUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042713
          miRBase ID: oni-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-23b
          Stem-loop Accession # MI0040292
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049526
          miRBase ID: pha-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-23b
          Stem-loop Accession # MI0034903
          Stem-loop Sequence
          GGGUUCCUGGCGUGCUGAUUUGUGACUUAUGAUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042924
          miRBase ID: pny-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-23b
          Stem-loop Accession # MI0007592
          Stem-loop Sequence
          CUCAGGUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCACGACCUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006165
          miRBase ID: mml-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-23
          Stem-loop Accession # MI0013787
          Stem-loop Sequence
          UUUGGGUUCCUGGCAUGAUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAGCCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014566
          miRBase ID: tgu-miR-23-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-23b
          Stem-loop Accession # MI0038304
          Stem-loop Sequence
          UGGGUUCCUGGCAUGCUGAUUUGUGAGUUAAGAUUAAAAUCACAUUGCCAGGGAUUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046549
          miRBase ID: xla-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-23b
          Stem-loop Accession # MI0004913
          Stem-loop Sequence
          CCGGUGUGGCUGUUUGGGUUCCUGGCAUGCUGAUUUGUGAGUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACACAACCAUGUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003673
          miRBase ID: xtr-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306184_pri
          TaqMan™ Pri-miRNA Assay : Rn03465744_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10711
          Ambion® Pre-miR™ miRNA Precursor : PM10711
          TaqMan™ MicroRNA Assay : 000400
          mirVana® miRNA inhibitor : MH10711
          TaqMan™ Advanced miRNA Assay : rno478602_mir
          TaqMan™ Advanced miRNA Assay : mmu478602_mir
          TaqMan™ Advanced miRNA Assay : 478602_mir


          miRBase Alias:

          Human: hsa-miR-23b-3p (v21)
          Human: hsa-miR-23b (v17)
          Mouse: mmu-miR-23b (v17)
          Rat: rno-miR-23b (v18)
          Aeromonas caviae Ae398: aca-miR-23b (v18)
          Gallus gallus: gga-miR-23b (v19)
          Macaca mulatta: mml-miR-23b (v19)
          Monodelphis domestica: mdo-miR-23b (v19)
          Trabulsiella guamensis ATCC 49490: tgu-miR-23 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline